Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218982145-218982386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEV
Entrez Gene ID
54738
Gene Name
FEV, ETS transcription factor
Gene Aliases
HSRNAFEV, PET-1
UniGene
Hs.234759
Gene Chromosome Location
Chr.2: 218981087-218985657 on Build GRCh38
Transcripts
NM_017521.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4684378 c.200C>T p.A67V Chr.2: 218982184-218982184 on Build GRCh38
5809261 c.227A>T p.K76M Chr.2: 218982157-218982157 on Build GRCh38
4091558 c.211G>T p.G71C Chr.2: 218982173-218982173 on Build GRCh38
4534119 c.204G>A p.W68* Chr.2: 218982180-218982180 on Build GRCh38
3838615 c.220G>C p.E74Q Chr.2: 218982164-218982164 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891762005 Chr.2: 218982352-218982352 on Build GRCh38
rs922025030 Chr.2: 218982304-218982304 on Build GRCh38
rs931811227 Chr.2: 218982357-218982357 on Build GRCh38
rs991867798 Chr.2: 218982362-218982362 on Build GRCh38
rs754550407 Chr.2: 218982287-218982287 on Build GRCh38
rs1035785950 Chr.2: 218982327-218982327 on Build GRCh38
rs561811754 Chr.2: 218982267-218982267 on Build GRCh38
rs573116414 Chr.2: 218982236-218982236 on Build GRCh38
rs452985 Chr.2: 218982293-218982293 on Build GRCh38
rs1008910596 Chr.2: 218982354-218982354 on Build GRCh38
rs759630260 Chr.2: 218982247-218982247 on Build GRCh38
rs774493524 Chr.2: 218982265-218982265 on Build GRCh38
rs1053631304 Chr.2: 218982343-218982343 on Build GRCh38
rs898052392 Chr.2: 218982318-218982319 on Build GRCh38
rs1006509151 Chr.2: 218982310-218982310 on Build GRCh38
rs748650671 Chr.2: 218982198-218982198 on Build GRCh38
rs760382278 Chr.2: 218982271-218982271 on Build GRCh38
rs917489343 Chr.2: 218982166-218982166 on Build GRCh38
rs779455782 Chr.2: 218982196-218982196 on Build GRCh38
rs748683998 Chr.2: 218982192-218982192 on Build GRCh38
rs753480146 Chr.2: 218982279-218982279 on Build GRCh38
rs765894103 Chr.2: 218982272-218982272 on Build GRCh38
rs1040433308 Chr.2: 218982358-218982358 on Build GRCh38
rs772468410 Chr.2: 218982201-218982201 on Build GRCh38
rs1016907524 Chr.2: 218982316-218982316 on Build GRCh38
rs767391303 Chr.2: 218982268-218982268 on Build GRCh38
rs776776811 Chr.2: 218982244-218982244 on Build GRCh38
rs755493413 Chr.2: 218982195-218982195 on Build GRCh38
rs750147210 Chr.2: 218982269-218982269 on Build GRCh38
rs994083594 Chr.2: 218982322-218982322 on Build GRCh38
rs200783809 Chr.2: 218982261-218982261 on Build GRCh38
rs369350686 Chr.2: 218982299-218982299 on Build GRCh38

View Full Product Details