Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 47347693-47347948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB13
Entrez Gene ID
124989
Gene Name
EF-hand calcium binding domain 13
Gene Aliases
C17orf57
UniGene
Hs.463303
Gene Chromosome Location
Chr.17: 47323961-47441312 on Build GRCh38
Transcripts
NM_001195192.1 NM_152347.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5426256 c.631C>T p.R211* Chr.17: 47347921-47347921 on Build GRCh38
383907 c.613A>G p.I205V Chr.17: 47347903-47347903 on Build GRCh38
1750099 c.652G>A p.D218N Chr.17: 47347942-47347942 on Build GRCh38
4553928 c.605G>C p.R202T Chr.17: 47347895-47347895 on Build GRCh38
6882795 c.560G>A p.G187D Chr.17: 47347850-47347850 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752425443 Chr.17: 47347774-47347774 on Build GRCh38
rs377511631 Chr.17: 47347796-47347796 on Build GRCh38
rs553472420 Chr.17: 47347857-47347857 on Build GRCh38
rs754342070 Chr.17: 47347798-47347798 on Build GRCh38
rs755332890 Chr.17: 47347800-47347800 on Build GRCh38
rs767013324 Chr.17: 47347916-47347916 on Build GRCh38
rs780477958 Chr.17: 47347797-47347797 on Build GRCh38
rs552280698 Chr.17: 47347723-47347723 on Build GRCh38
rs751236063 Chr.17: 47347898-47347898 on Build GRCh38
rs554271269 Chr.17: 47347755-47347755 on Build GRCh38
rs201799724 Chr.17: 47347903-47347903 on Build GRCh38
rs201299500 Chr.17: 47347843-47347843 on Build GRCh38
rs763904554 Chr.17: 47347896-47347896 on Build GRCh38
rs776616673 Chr.17: 47347765-47347765 on Build GRCh38
rs150498027 Chr.17: 47347814-47347814 on Build GRCh38
rs71377306 Chr.17: 47347921-47347921 on Build GRCh38
rs577139493 Chr.17: 47347815-47347815 on Build GRCh38
rs1002673460 Chr.17: 47347727-47347727 on Build GRCh38
rs534660403 Chr.17: 47347751-47347751 on Build GRCh38
rs751043940 Chr.17: 47347793-47347793 on Build GRCh38
rs779257942 Chr.17: 47347922-47347922 on Build GRCh38
rs748383264 Chr.17: 47347810-47347810 on Build GRCh38
rs912832963 Chr.17: 47347803-47347803 on Build GRCh38
rs114861884 Chr.17: 47347788-47347788 on Build GRCh38
rs779288324 Chr.17: 47347809-47347809 on Build GRCh38
rs762698524 Chr.17: 47347883-47347883 on Build GRCh38
rs149684195 Chr.17: 47347844-47347844 on Build GRCh38
rs759297917 Chr.17: 47347766-47347766 on Build GRCh38
rs775366829 Chr.17: 47347873-47347873 on Build GRCh38
rs200266257 Chr.17: 47347918-47347918 on Build GRCh38
rs3851808 Chr.17: 47347778-47347778 on Build GRCh38
rs769499375 Chr.17: 47347867-47347867 on Build GRCh38
rs747147953 Chr.17: 47347820-47347820 on Build GRCh38
rs761270201 Chr.17: 47347899-47347899 on Build GRCh38
rs765050657 Chr.17: 47347767-47347767 on Build GRCh38
rs1036202895 Chr.17: 47347743-47347743 on Build GRCh38
rs7206971 Chr.17: 47347749-47347749 on Build GRCh38

View Full Product Details