Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 78013467-78013696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB4
Entrez Gene ID
11080
Gene Name
DnaJ heat shock protein family (Hsp40) member B4
Gene Aliases
DNAJW, DjB4, HLJ1
UniGene
Hs.13852
Gene Chromosome Location
Chr.1: 77979158-78017964 on Build GRCh38
Transcripts
NM_001317099.1 NM_001317100.1 NM_001317101.1 NM_001317102.1 NM_001317103.1 NM_007034.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6127554 c.644C>T p.T215I Chr.1: 78013483-78013483 on Build GRCh38
4396128 c.653G>A p.R218K Chr.1: 78013492-78013492 on Build GRCh38
6647121 c.776G>A p.R259Q Chr.1: 78013615-78013615 on Build GRCh38
6647120 c.658G>T p.G220* Chr.1: 78013497-78013497 on Build GRCh38
6647123 c.727T>G p.F243V Chr.1: 78013566-78013566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751877573 Chr.1: 78013530-78013530 on Build GRCh38
rs1013057710 Chr.1: 78013518-78013518 on Build GRCh38
rs774863543 Chr.1: 78013630-78013630 on Build GRCh38
rs770572389 Chr.1: 78013581-78013581 on Build GRCh38
rs746729437 Chr.1: 78013573-78013573 on Build GRCh38
rs200647412 Chr.1: 78013663-78013663 on Build GRCh38
rs778941840 Chr.1: 78013555-78013555 on Build GRCh38
rs775667040 Chr.1: 78013520-78013520 on Build GRCh38
rs764607863 Chr.1: 78013615-78013615 on Build GRCh38
rs983156640 Chr.1: 78013604-78013604 on Build GRCh38
rs745404922 Chr.1: 78013591-78013591 on Build GRCh38
rs777392863 Chr.1: 78013570-78013570 on Build GRCh38
rs566548207 Chr.1: 78013603-78013603 on Build GRCh38
rs763438687 Chr.1: 78013525-78013525 on Build GRCh38
rs776099937 Chr.1: 78013600-78013600 on Build GRCh38
rs367875634 Chr.1: 78013504-78013504 on Build GRCh38
rs763384976 Chr.1: 78013614-78013614 on Build GRCh38
rs758240279 Chr.1: 78013563-78013563 on Build GRCh38
rs11589750 Chr.1: 78013597-78013597 on Build GRCh38
rs750533869 Chr.1: 78013535-78013535 on Build GRCh38
rs145798922 Chr.1: 78013534-78013534 on Build GRCh38
rs773054179 Chr.1: 78013661-78013661 on Build GRCh38
rs1003071459 Chr.1: 78013660-78013660 on Build GRCh38
rs764376696 Chr.1: 78013529-78013529 on Build GRCh38
rs747864591 Chr.1: 78013558-78013558 on Build GRCh38
rs754826655 Chr.1: 78013542-78013542 on Build GRCh38
rs368881437 Chr.1: 78013531-78013531 on Build GRCh38
rs775945067 Chr.1: 78013587-78013587 on Build GRCh38

View Full Product Details