Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4972201-4972474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMTA2
Entrez Gene ID
23125
Gene Name
calmodulin binding transcription activator 2
Gene Aliases
-
UniGene
Hs.632242
Gene Chromosome Location
Chr.17: 4967992-4988297 on Build GRCh38
Transcripts
NM_001171166.1 NM_001171167.1 NM_001171168.1 NM_015099.3 XM_006721478.3 XM_006721481.3 XM_006721482.3 XM_011523746.2 XM_011523747.2 XM_011523748.2 XM_011523749.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4998539 c.2778G>C p.E926D Chr.17: 4972268-4972268 on Build GRCh38
5475945 c.2667T>C p.P889P Chr.17: 4972442-4972442 on Build GRCh38
4484309 c.2796C>T p.S932S Chr.17: 4972313-4972313 on Build GRCh38
5475946 c.2598T>C p.P866P Chr.17: 4972442-4972442 on Build GRCh38
6002926 c.2725C>A p.L909I Chr.17: 4972321-4972321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765604347 Chr.17: 4972326-4972326 on Build GRCh38
rs150571966 Chr.17: 4972347-4972347 on Build GRCh38
rs755045273 Chr.17: 4972443-4972443 on Build GRCh38
rs779062904 Chr.17: 4972446-4972446 on Build GRCh38
rs745699429 Chr.17: 4972364-4972364 on Build GRCh38
rs765769746 Chr.17: 4972409-4972409 on Build GRCh38
rs774625299 Chr.17: 4972406-4972406 on Build GRCh38
rs779694510 Chr.17: 4972450-4972450 on Build GRCh38
rs754229322 Chr.17: 4972235-4972235 on Build GRCh38
rs749213515 Chr.17: 4972346-4972346 on Build GRCh38
rs373003229 Chr.17: 4972255-4972255 on Build GRCh38
rs1053449625 Chr.17: 4972303-4972303 on Build GRCh38
rs961833349 Chr.17: 4972402-4972402 on Build GRCh38
rs780204153 Chr.17: 4972332-4972332 on Build GRCh38
rs758145052 Chr.17: 4972448-4972448 on Build GRCh38
rs544422438 Chr.17: 4972236-4972236 on Build GRCh38
rs149983655 Chr.17: 4972407-4972407 on Build GRCh38
rs1056414006 Chr.17: 4972323-4972323 on Build GRCh38
rs777628126 Chr.17: 4972272-4972272 on Build GRCh38
rs778829811 Chr.17: 4972350-4972350 on Build GRCh38
rs76796952 Chr.17: 4972245-4972245 on Build GRCh38
rs763363113 Chr.17: 4972426-4972426 on Build GRCh38
rs16942615 Chr.17: 4972333-4972333 on Build GRCh38
rs756902000 Chr.17: 4972442-4972443 on Build GRCh38
rs147321617 Chr.17: 4972265-4972265 on Build GRCh38
rs74916567 Chr.17: 4972242-4972242 on Build GRCh38
rs760172832 Chr.17: 4972320-4972320 on Build GRCh38
rs943912192 Chr.17: 4972240-4972240 on Build GRCh38
rs751732857 Chr.17: 4972433-4972433 on Build GRCh38
rs774107039 Chr.17: 4972283-4972283 on Build GRCh38
rs142999940 Chr.17: 4972264-4972264 on Build GRCh38
rs1018563190 Chr.17: 4972429-4972429 on Build GRCh38
rs745547658 Chr.17: 4972443-4972444 on Build GRCh38
rs78912625 Chr.17: 4972403-4972403 on Build GRCh38
rs779053378 Chr.17: 4972262-4972262 on Build GRCh38
rs771847281 Chr.17: 4972377-4972377 on Build GRCh38
rs199714272 Chr.17: 4972342-4972342 on Build GRCh38
rs770853831 Chr.17: 4972282-4972282 on Build GRCh38
rs141604881 Chr.17: 4972341-4972341 on Build GRCh38
rs778381439 Chr.17: 4972443-4972443 on Build GRCh38
rs238230 Chr.17: 4972271-4972271 on Build GRCh38
rs771902140 Chr.17: 4972308-4972308 on Build GRCh38
rs144794944 Chr.17: 4972310-4972310 on Build GRCh38
rs764651342 Chr.17: 4972324-4972324 on Build GRCh38
rs986659498 Chr.17: 4972421-4972421 on Build GRCh38
rs183231630 Chr.17: 4972304-4972304 on Build GRCh38
rs773559133 Chr.17: 4972413-4972413 on Build GRCh38
rs566382896 Chr.17: 4972431-4972431 on Build GRCh38
rs201333256 Chr.17: 4972396-4972396 on Build GRCh38
rs768060485 Chr.17: 4972400-4972400 on Build GRCh38
rs34249300 Chr.17: 4972263-4972263 on Build GRCh38
rs200654310 Chr.17: 4972325-4972325 on Build GRCh38
rs750831335 Chr.17: 4972327-4972327 on Build GRCh38
rs749052822 Chr.17: 4972279-4972279 on Build GRCh38
rs200839594 Chr.17: 4972444-4972444 on Build GRCh38
rs565049159 Chr.17: 4972328-4972328 on Build GRCh38
rs367576965 Chr.17: 4972267-4972267 on Build GRCh38

View Full Product Details