Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9734075-9734320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRPF1
Entrez Gene ID
7862
Gene Name
bromodomain and PHD finger containing 1
Gene Aliases
BR140
UniGene
Hs.1004
Gene Chromosome Location
Chr.3: 9731729-9748015 on Build GRCh38
Transcripts
NM_001003694.1 NM_001319049.1 NM_001319050.1 NM_004634.2 XM_005265449.1 XM_005265450.1 XM_005265452.1 XM_011534101.1 XM_011534102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6612901 c.33C>T p.C11C Chr.3: 9734173-9734173 on Build GRCh38
7197825 c.115G>A p.E39K Chr.3: 9734255-9734255 on Build GRCh38
1425790 c.47C>T p.A16V Chr.3: 9734187-9734187 on Build GRCh38
5452550 c.124C>G p.L42V Chr.3: 9734264-9734264 on Build GRCh38
4851130 c.158C>A p.P53Q Chr.3: 9734298-9734298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748562779 Chr.3: 9734224-9734224 on Build GRCh38
rs1007331876 Chr.3: 9734282-9734282 on Build GRCh38
rs774820384 Chr.3: 9734286-9734286 on Build GRCh38
rs537812311 Chr.3: 9734175-9734175 on Build GRCh38
rs200084177 Chr.3: 9734098-9734098 on Build GRCh38
rs745792490 Chr.3: 9734200-9734200 on Build GRCh38
rs770024332 Chr.3: 9734226-9734226 on Build GRCh38
rs763886067 Chr.3: 9734295-9734295 on Build GRCh38
rs887553174 Chr.3: 9734158-9734158 on Build GRCh38
rs151300467 Chr.3: 9734161-9734161 on Build GRCh38
rs907077759 Chr.3: 9734124-9734124 on Build GRCh38
rs750976982 Chr.3: 9734178-9734178 on Build GRCh38
rs777004522 Chr.3: 9734218-9734218 on Build GRCh38
rs762336566 Chr.3: 9734118-9734118 on Build GRCh38
rs74450417 Chr.3: 9734159-9734159 on Build GRCh38
rs116359925 Chr.3: 9734203-9734203 on Build GRCh38
rs374404519 Chr.3: 9734107-9734107 on Build GRCh38
rs753506438 Chr.3: 9734296-9734296 on Build GRCh38
rs557815695 Chr.3: 9734115-9734115 on Build GRCh38
rs146653692 Chr.3: 9734281-9734281 on Build GRCh38
rs752408281 Chr.3: 9734155-9734155 on Build GRCh38
rs551278897 Chr.3: 9734104-9734104 on Build GRCh38
rs761763071 Chr.3: 9734298-9734298 on Build GRCh38
rs757099971 Chr.3: 9734188-9734188 on Build GRCh38
rs765682102 Chr.3: 9734133-9734133 on Build GRCh38
rs768828309 Chr.3: 9734212-9734212 on Build GRCh38
rs773570041 Chr.3: 9734139-9734139 on Build GRCh38
rs759125488 Chr.3: 9734146-9734146 on Build GRCh38
rs749634353 Chr.3: 9734136-9734137 on Build GRCh38
rs753641137 Chr.3: 9734184-9734184 on Build GRCh38
rs868370823 Chr.3: 9734183-9734183 on Build GRCh38
rs954283319 Chr.3: 9734230-9734230 on Build GRCh38
rs779936383 Chr.3: 9734204-9734204 on Build GRCh38
rs1057519517 Chr.3: 9734244-9734245 on Build GRCh38
rs773828051 Chr.3: 9734245-9734245 on Build GRCh38
rs763310765 Chr.3: 9734248-9734248 on Build GRCh38

View Full Product Details