Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57469540-57469769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLI1
Entrez Gene ID
2735
Gene Name
GLI family zinc finger 1
Gene Aliases
GLI
UniGene
Hs.632702
Gene Chromosome Location
Chr.12: 57460135-57472268 on Build GRCh38
Transcripts
NM_001160045.1 NM_001167609.1 NM_005269.2 XM_011538189.2 XM_011538190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
244708 c.1574C>A p.T525N Chr.12: 57469696-57469696 on Build GRCh38
4209724 c.1490G>A p.R497H Chr.12: 57469612-57469612 on Build GRCh38
6977425 c.1529G>A p.R510Q Chr.12: 57469651-57469651 on Build GRCh38
1677114 c.1432C>T p.L478F Chr.12: 57469554-57469554 on Build GRCh38
6952013 c.1522C>T p.Q508* Chr.12: 57469644-57469644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566564028 Chr.12: 57469656-57469656 on Build GRCh38
rs756280054 Chr.12: 57469697-57469697 on Build GRCh38
rs761540711 Chr.12: 57469712-57469712 on Build GRCh38
rs375428680 Chr.12: 57469665-57469665 on Build GRCh38
rs768365587 Chr.12: 57469708-57469708 on Build GRCh38
rs781683841 Chr.12: 57469666-57469666 on Build GRCh38
rs765045779 Chr.12: 57469715-57469715 on Build GRCh38
rs573996360 Chr.12: 57469661-57469661 on Build GRCh38
rs781723374 Chr.12: 57469601-57469601 on Build GRCh38
rs746661249 Chr.12: 57469705-57469705 on Build GRCh38
rs74607574 Chr.12: 57469682-57469682 on Build GRCh38
rs753690500 Chr.12: 57469639-57469639 on Build GRCh38
rs775171324 Chr.12: 57469612-57469612 on Build GRCh38
rs139792497 Chr.12: 57469608-57469608 on Build GRCh38
rs769033058 Chr.12: 57469594-57469594 on Build GRCh38
rs760289179 Chr.12: 57469627-57469627 on Build GRCh38
rs551972267 Chr.12: 57469652-57469652 on Build GRCh38
rs185679191 Chr.12: 57469593-57469593 on Build GRCh38
rs113261025 Chr.12: 57469679-57469679 on Build GRCh38
rs774916232 Chr.12: 57469698-57469698 on Build GRCh38
rs759027001 Chr.12: 57469609-57469609 on Build GRCh38
rs201902084 Chr.12: 57469607-57469607 on Build GRCh38
rs368186732 Chr.12: 57469588-57469588 on Build GRCh38
rs533875222 Chr.12: 57469611-57469611 on Build GRCh38
rs199949777 Chr.12: 57469568-57469568 on Build GRCh38
rs890152411 Chr.12: 57469626-57469626 on Build GRCh38
rs990717142 Chr.12: 57469684-57469684 on Build GRCh38
rs778951240 Chr.12: 57469576-57469576 on Build GRCh38
rs780272131 Chr.12: 57469590-57469590 on Build GRCh38
rs963099490 Chr.12: 57469659-57469659 on Build GRCh38
rs1030336260 Chr.12: 57469731-57469731 on Build GRCh38
rs375299184 Chr.12: 57469723-57469723 on Build GRCh38
rs776271855 Chr.12: 57469711-57469711 on Build GRCh38
rs761735108 Chr.12: 57469649-57469649 on Build GRCh38
rs1036861417 Chr.12: 57469728-57469728 on Build GRCh38
rs149817893 Chr.12: 57469650-57469650 on Build GRCh38
rs748647850 Chr.12: 57469604-57469604 on Build GRCh38
rs572499706 Chr.12: 57469655-57469655 on Build GRCh38
rs778986643 Chr.12: 57469710-57469711 on Build GRCh38
rs147931475 Chr.12: 57469651-57469651 on Build GRCh38
rs763858681 Chr.12: 57469629-57469629 on Build GRCh38
rs757858450 Chr.12: 57469619-57469619 on Build GRCh38
rs12578516 Chr.12: 57469605-57469605 on Build GRCh38
rs750584946 Chr.12: 57469579-57469579 on Build GRCh38
rs372330353 Chr.12: 57469637-57469637 on Build GRCh38
rs866686999 Chr.12: 57469638-57469638 on Build GRCh38

View Full Product Details