Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 34365156-34365381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPI
Entrez Gene ID
2821
Gene Name
glucose-6-phosphate isomerase
Gene Aliases
AMF, GNPI, NLK, PGI, PHI, SA-36, SA36
UniGene
Hs.466471
Gene Chromosome Location
Chr.19: 34359592-34402413 on Build GRCh38
Transcripts
NM_000175.3 NM_001184722.1 NM_001289789.1 NM_001289790.1 XM_005258764.2 XM_006723148.2 XM_011526754.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6408558 c.115G>A p.A39T Chr.19: 34365264-34365264 on Build GRCh38
7251519 c.170G>C p.R57P Chr.19: 34365319-34365319 on Build GRCh38
5833700 c.133_134insG p.D46fs*82 Chr.19: 34365282-34365283 on Build GRCh38
7251520 c.53G>C p.R18P Chr.19: 34365319-34365319 on Build GRCh38
1325333 c.45A>G p.Q15Q Chr.19: 34365311-34365311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773642615 Chr.19: 34365223-34365223 on Build GRCh38
rs990168476 Chr.19: 34365335-34365335 on Build GRCh38
rs370484673 Chr.19: 34365175-34365175 on Build GRCh38
rs777217086 Chr.19: 34365239-34365242 on Build GRCh38
rs755993078 Chr.19: 34365251-34365251 on Build GRCh38
rs572523544 Chr.19: 34365311-34365311 on Build GRCh38
rs778943515 Chr.19: 34365253-34365253 on Build GRCh38
rs777848549 Chr.19: 34365259-34365259 on Build GRCh38
rs751093058 Chr.19: 34365278-34365278 on Build GRCh38
rs771058879 Chr.19: 34365329-34365329 on Build GRCh38
rs781623690 Chr.19: 34365333-34365333 on Build GRCh38
rs944710510 Chr.19: 34365197-34365197 on Build GRCh38
rs11549001 Chr.19: 34365332-34365332 on Build GRCh38
rs761450928 Chr.19: 34365282-34365282 on Build GRCh38
rs750236148 Chr.19: 34365247-34365247 on Build GRCh38
rs772206226 Chr.19: 34365220-34365220 on Build GRCh38
rs781329755 Chr.19: 34365241-34365241 on Build GRCh38
rs535827434 Chr.19: 34365233-34365233 on Build GRCh38
rs567488016 Chr.19: 34365200-34365200 on Build GRCh38
rs977657435 Chr.19: 34365236-34365236 on Build GRCh38
rs777192502 Chr.19: 34365265-34365265 on Build GRCh38
rs752455390 Chr.19: 34365232-34365232 on Build GRCh38
rs550848083 Chr.19: 34365195-34365195 on Build GRCh38
rs781164783 Chr.19: 34365215-34365215 on Build GRCh38
rs770211568 Chr.19: 34365271-34365271 on Build GRCh38
rs915331981 Chr.19: 34365190-34365190 on Build GRCh38
rs267606852 Chr.19: 34365280-34365280 on Build GRCh38
rs746395554 Chr.19: 34365351-34365351 on Build GRCh38
rs776955537 Chr.19: 34365229-34365229 on Build GRCh38
rs888216812 Chr.19: 34365224-34365224 on Build GRCh38
rs915385692 Chr.19: 34365243-34365243 on Build GRCh38
rs764796276 Chr.19: 34365231-34365231 on Build GRCh38
rs774942905 Chr.19: 34365272-34365272 on Build GRCh38
rs762697901 Chr.19: 34365275-34365275 on Build GRCh38
rs369198577 Chr.19: 34365302-34365302 on Build GRCh38
rs766677031 Chr.19: 34365227-34365227 on Build GRCh38
rs768741049 Chr.19: 34365217-34365217 on Build GRCh38
rs910513006 Chr.19: 34365196-34365196 on Build GRCh38
rs759987877 Chr.19: 34365230-34365230 on Build GRCh38
rs750387485 Chr.19: 34365290-34365290 on Build GRCh38
rs151025852 Chr.19: 34365220-34365221 on Build GRCh38
rs767547881 Chr.19: 34365288-34365288 on Build GRCh38
rs374135310 Chr.19: 34365234-34365234 on Build GRCh38
rs763277247 Chr.19: 34365359-34365359 on Build GRCh38
rs747137710 Chr.19: 34365261-34365261 on Build GRCh38
rs752227961 Chr.19: 34365223-34365223 on Build GRCh38
rs774699183 Chr.19: 34365219-34365219 on Build GRCh38
rs755798489 Chr.19: 34365293-34365293 on Build GRCh38
rs901086822 Chr.19: 34365211-34365211 on Build GRCh38
rs1046891815 Chr.19: 34365222-34365222 on Build GRCh38
rs761197420 Chr.19: 34365226-34365226 on Build GRCh38
rs946761670 Chr.19: 34365193-34365193 on Build GRCh38
rs368450858 Chr.19: 34365356-34365356 on Build GRCh38
rs148025432 Chr.19: 34365353-34365353 on Build GRCh38
rs137853586 Chr.19: 34365325-34365325 on Build GRCh38
rs749474404 Chr.19: 34365216-34365216 on Build GRCh38
rs759810325 Chr.19: 34365268-34365268 on Build GRCh38
rs758060545 Chr.19: 34365223-34365226 on Build GRCh38
rs372044576 Chr.19: 34365321-34365321 on Build GRCh38
rs552574934 Chr.19: 34365258-34365258 on Build GRCh38
rs771393881 Chr.19: 34365264-34365264 on Build GRCh38
rs747084719 Chr.19: 34365328-34365328 on Build GRCh38
rs8191359 Chr.19: 34365209-34365209 on Build GRCh38
rs763650536 Chr.19: 34365276-34365276 on Build GRCh38

View Full Product Details