Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92500654-92500881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPP
Entrez Gene ID
401541
Gene Name
centromere protein P
Gene Aliases
CENP-P
UniGene
Hs.713775
Gene Chromosome Location
Chr.9: 92325457-92615164 on Build GRCh38
Transcripts
NM_001012267.2 NM_001286969.1 NM_001197295.1 NM_001197296.1 NM_001393.3

Gene Symbol
ECM2
Entrez Gene ID
1842
Gene Name
extracellular matrix protein 2
Gene Aliases
-
UniGene
Hs.117060
Gene Chromosome Location
Chr.9: 92493547-92536092 on Build GRCh38
Transcripts
NM_001012267.2 NM_001286969.1 NM_001197295.1 NM_001197296.1 NM_001393.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1463597 c.1768G>T p.E590* Chr.9: 92500824-92500824 on Build GRCh38
1463595 c.1799C>G p.S600C Chr.9: 92500793-92500793 on Build GRCh38
3927221 c.1720G>A p.D574N Chr.9: 92500872-92500872 on Build GRCh38
1463596 c.1834G>T p.E612* Chr.9: 92500824-92500824 on Build GRCh38
3908891 c.1798C>T p.R600C Chr.9: 92500860-92500860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200394678 Chr.9: 92500727-92500727 on Build GRCh38
rs918395072 Chr.9: 92500716-92500716 on Build GRCh38
rs778464812 Chr.9: 92500826-92500826 on Build GRCh38
rs776930712 Chr.9: 92500757-92500757 on Build GRCh38
rs942352232 Chr.9: 92500843-92500843 on Build GRCh38
rs140117576 Chr.9: 92500723-92500723 on Build GRCh38
rs770032853 Chr.9: 92500762-92500762 on Build GRCh38
rs1039336331 Chr.9: 92500846-92500846 on Build GRCh38
rs570789332 Chr.9: 92500859-92500859 on Build GRCh38
rs780638094 Chr.9: 92500857-92500857 on Build GRCh38
rs370627896 Chr.9: 92500729-92500729 on Build GRCh38
rs761774737 Chr.9: 92500758-92500758 on Build GRCh38
rs138868045 Chr.9: 92500807-92500807 on Build GRCh38
rs774424324 Chr.9: 92500766-92500766 on Build GRCh38
rs151070730 Chr.9: 92500728-92500728 on Build GRCh38
rs752381563 Chr.9: 92500691-92500691 on Build GRCh38
rs746618975 Chr.9: 92500733-92500735 on Build GRCh38
rs756731840 Chr.9: 92500825-92500825 on Build GRCh38
rs775779497 Chr.9: 92500735-92500735 on Build GRCh38
rs750921084 Chr.9: 92500839-92500839 on Build GRCh38
rs150249574 Chr.9: 92500788-92500788 on Build GRCh38
rs752368753 Chr.9: 92500787-92500787 on Build GRCh38
rs758832338 Chr.9: 92500855-92500855 on Build GRCh38
rs764243970 Chr.9: 92500746-92500746 on Build GRCh38
rs201745324 Chr.9: 92500847-92500847 on Build GRCh38
rs776211051 Chr.9: 92500860-92500860 on Build GRCh38
rs982943174 Chr.9: 92500819-92500822 on Build GRCh38
rs763606251 Chr.9: 92500803-92500803 on Build GRCh38
rs770551588 Chr.9: 92500848-92500850 on Build GRCh38
rs767486657 Chr.9: 92500690-92500690 on Build GRCh38
rs748708631 Chr.9: 92500705-92500705 on Build GRCh38
rs186648835 Chr.9: 92500831-92500831 on Build GRCh38
rs547551173 Chr.9: 92500785-92500785 on Build GRCh38
rs140976209 Chr.9: 92500776-92500776 on Build GRCh38
rs767576492 Chr.9: 92500780-92500780 on Build GRCh38
rs755665680 Chr.9: 92500703-92500703 on Build GRCh38

View Full Product Details