Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9474606-9474847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IAH1
Entrez Gene ID
285148
Gene Name
isoamyl acetate-hydrolyzing esterase 1 homolog
Gene Aliases
-
UniGene
Hs.667226
Gene Chromosome Location
Chr.2: 9473638-9512403 on Build GRCh38
Transcripts
NM_001039613.2 NM_001320859.1 NM_001320860.1 NM_001320863.1 XM_017003878.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6159327 c.49C>T p.R17C Chr.2: 9474615-9474615 on Build GRCh38
5397581 c.67G>A p.D23N Chr.2: 9474633-9474633 on Build GRCh38
6381867 c.77C>A p.T26N Chr.2: 9474643-9474643 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs116238865 Chr.2: 9474784-9474784 on Build GRCh38
rs758643719 Chr.2: 9474632-9474632 on Build GRCh38
rs563636372 Chr.2: 9474660-9474660 on Build GRCh38
rs768298085 Chr.2: 9474693-9474693 on Build GRCh38
rs751767561 Chr.2: 9474638-9474638 on Build GRCh38
rs12996760 Chr.2: 9474688-9474688 on Build GRCh38
rs899171093 Chr.2: 9474825-9474825 on Build GRCh38
rs543844705 Chr.2: 9474652-9474652 on Build GRCh38
rs1006837196 Chr.2: 9474757-9474757 on Build GRCh38
rs745696507 Chr.2: 9474671-9474671 on Build GRCh38
rs767481075 Chr.2: 9474654-9474673 on Build GRCh38
rs946104647 Chr.2: 9474799-9474799 on Build GRCh38
rs965219148 Chr.2: 9474800-9474800 on Build GRCh38
rs771721482 Chr.2: 9474681-9474681 on Build GRCh38
rs776125989 Chr.2: 9474697-9474697 on Build GRCh38
rs1010498065 Chr.2: 9474831-9474831 on Build GRCh38
rs868812817 Chr.2: 9474826-9474826 on Build GRCh38
rs1038076304 Chr.2: 9474814-9474814 on Build GRCh38
rs762377321 Chr.2: 9474623-9474623 on Build GRCh38
rs924772695 Chr.2: 9474707-9474707 on Build GRCh38
rs781479118 Chr.2: 9474651-9474651 on Build GRCh38
rs1026395408 Chr.2: 9474829-9474829 on Build GRCh38
rs889387352 Chr.2: 9474795-9474795 on Build GRCh38
rs775337108 Chr.2: 9474683-9474683 on Build GRCh38
rs571487863 Chr.2: 9474629-9474629 on Build GRCh38
rs1042163716 Chr.2: 9474705-9474705 on Build GRCh38
rs933512535 Chr.2: 9474774-9474774 on Build GRCh38
rs765667385 Chr.2: 9474624-9474624 on Build GRCh38
rs889697855 Chr.2: 9474741-9474741 on Build GRCh38
rs780355850 Chr.2: 9474635-9474635 on Build GRCh38
rs1007826329 Chr.2: 9474801-9474801 on Build GRCh38
rs914926790 Chr.2: 9474649-9474649 on Build GRCh38
rs755202985 Chr.2: 9474640-9474640 on Build GRCh38

View Full Product Details