Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50750005-50750242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYLD
Entrez Gene ID
1540
Gene Name
CYLD lysine 63 deubiquitinase
Gene Aliases
BRSS, CDMT, CYLD1, CYLDI, EAC, MFT, MFT1, SBS, TEM, USPL2
UniGene
Hs.578973
Gene Chromosome Location
Chr.16: 50742026-50801935 on Build GRCh38
Transcripts
NM_001042355.1 NM_001042412.1 NM_015247.2 XM_005255812.2 XM_006721149.1 XM_011522907.2 XM_011522908.1 XM_011522911.2 XM_017022977.1 XM_017022978.1 XM_017022979.1 XM_017022980.1 XM_017022981.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
558068 c.341A>C p.K114T Chr.16: 50750039-50750039 on Build GRCh38
471801 c.388C>A p.P130T Chr.16: 50750086-50750086 on Build GRCh38
6354458 c.429T>A p.P143P Chr.16: 50750127-50750127 on Build GRCh38
4411773 c.472G>A p.V158I Chr.16: 50750170-50750170 on Build GRCh38
280251 c.478G>A p.G160R Chr.16: 50750176-50750176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758921558 Chr.16: 50750084-50750084 on Build GRCh38
rs778620114 Chr.16: 50750131-50750131 on Build GRCh38
rs747924691 Chr.16: 50750138-50750138 on Build GRCh38
rs940350665 Chr.16: 50750137-50750137 on Build GRCh38
rs891941021 Chr.16: 50750180-50750180 on Build GRCh38
rs557031108 Chr.16: 50750175-50750175 on Build GRCh38
rs766661461 Chr.16: 50750055-50750055 on Build GRCh38
rs765653845 Chr.16: 50750079-50750079 on Build GRCh38
rs750019749 Chr.16: 50750092-50750092 on Build GRCh38
rs369238843 Chr.16: 50750076-50750076 on Build GRCh38
rs760665500 Chr.16: 50750033-50750033 on Build GRCh38
rs772062242 Chr.16: 50750152-50750152 on Build GRCh38
rs997651760 Chr.16: 50750073-50750073 on Build GRCh38
rs777538408 Chr.16: 50750154-50750154 on Build GRCh38
rs748843520 Chr.16: 50750116-50750116 on Build GRCh38
rs587778224 Chr.16: 50750042-50750042 on Build GRCh38
rs373896011 Chr.16: 50750199-50750199 on Build GRCh38
rs779807077 Chr.16: 50750107-50750107 on Build GRCh38
rs754799573 Chr.16: 50750126-50750126 on Build GRCh38
rs764687557 Chr.16: 50750086-50750086 on Build GRCh38
rs753265479 Chr.16: 50750080-50750080 on Build GRCh38
rs549332039 Chr.16: 50750093-50750093 on Build GRCh38
rs776692170 Chr.16: 50750070-50750070 on Build GRCh38

View Full Product Details