Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184164802-184165027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DVL3
Entrez Gene ID
1857
Gene Name
dishevelled segment polarity protein 3
Gene Aliases
DRS3
UniGene
Hs.388116
Gene Chromosome Location
Chr.3: 184155311-184173614 on Build GRCh38
Transcripts
NM_004423.3 XM_005247172.2 XM_011512513.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400867 c.583G>C p.D195H Chr.3: 184164915-184164915 on Build GRCh38
3734222 c.569T>C p.F190S Chr.3: 184164901-184164901 on Build GRCh38
4634949 c.594C>T p.T198T Chr.3: 184164926-184164926 on Build GRCh38
70551 c.503G>A p.R168Q Chr.3: 184164835-184164835 on Build GRCh38
1041923 c.496C>T p.R166W Chr.3: 184164828-184164828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs994645566 Chr.3: 184164915-184164915 on Build GRCh38
rs563164795 Chr.3: 184165000-184165000 on Build GRCh38
rs373951906 Chr.3: 184164981-184164981 on Build GRCh38
rs897261501 Chr.3: 184164859-184164859 on Build GRCh38
rs754141293 Chr.3: 184164881-184164881 on Build GRCh38
rs762153830 Chr.3: 184164970-184164970 on Build GRCh38
rs775506324 Chr.3: 184164956-184164956 on Build GRCh38
rs746606032 Chr.3: 184164828-184164828 on Build GRCh38
rs761533548 Chr.3: 184164832-184164832 on Build GRCh38
rs768431588 Chr.3: 184164829-184164829 on Build GRCh38
rs764329274 Chr.3: 184164871-184164871 on Build GRCh38
rs764200445 Chr.3: 184164961-184164961 on Build GRCh38
rs143197489 Chr.3: 184164905-184164905 on Build GRCh38
rs757532123 Chr.3: 184164890-184164890 on Build GRCh38
rs780967881 Chr.3: 184164920-184164920 on Build GRCh38
rs770860159 Chr.3: 184164955-184164955 on Build GRCh38
rs750881394 Chr.3: 184164895-184164895 on Build GRCh38
rs1035344371 Chr.3: 184164949-184164949 on Build GRCh38
rs149103009 Chr.3: 184164855-184164855 on Build GRCh38
rs369764519 Chr.3: 184164974-184164974 on Build GRCh38
rs369586306 Chr.3: 184164963-184164963 on Build GRCh38
rs372800702 Chr.3: 184164835-184164835 on Build GRCh38
rs747871249 Chr.3: 184164923-184164923 on Build GRCh38
rs752163954 Chr.3: 184165003-184165003 on Build GRCh38
rs762740960 Chr.3: 184164943-184164943 on Build GRCh38
rs769468812 Chr.3: 184164924-184164924 on Build GRCh38
rs750807417 Chr.3: 184164999-184164999 on Build GRCh38
rs202225705 Chr.3: 184164848-184164848 on Build GRCh38
rs779402594 Chr.3: 184164891-184164891 on Build GRCh38
rs768376544 Chr.3: 184164912-184164912 on Build GRCh38
rs754504377 Chr.3: 184165008-184165008 on Build GRCh38
rs1021896193 Chr.3: 184164998-184164998 on Build GRCh38
rs767570260 Chr.3: 184164854-184164854 on Build GRCh38
rs746552480 Chr.3: 184164910-184164910 on Build GRCh38
rs760485854 Chr.3: 184164957-184164957 on Build GRCh38
rs760819076 Chr.3: 184164867-184164867 on Build GRCh38
rs999040417 Chr.3: 184165005-184165005 on Build GRCh38
rs151229784 Chr.3: 184164925-184164925 on Build GRCh38
rs775396369 Chr.3: 184164911-184164911 on Build GRCh38
rs776529678 Chr.3: 184164831-184164831 on Build GRCh38

View Full Product Details