Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100124469-100124742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNPY4
Entrez Gene ID
245812
Gene Name
canopy FGF signaling regulator 4
Gene Aliases
PRAT4B
UniGene
Hs.632293
Gene Chromosome Location
Chr.7: 100119642-100125508 on Build GRCh38
Transcripts
NM_152755.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7277054 c.466-2A>G p.? Chr.7: 100124512-100124512 on Build GRCh38
3768608 c.517C>T p.H173Y Chr.7: 100124565-100124565 on Build GRCh38
4946516 c.511T>C p.Y171H Chr.7: 100124559-100124559 on Build GRCh38
1568769 c.513C>T p.Y171Y Chr.7: 100124561-100124561 on Build GRCh38
384112 c.574G>A p.A192T Chr.7: 100124622-100124622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019356159 Chr.7: 100124713-100124713 on Build GRCh38
rs373731246 Chr.7: 100124649-100124649 on Build GRCh38
rs748148095 Chr.7: 100124717-100124717 on Build GRCh38
rs745386990 Chr.7: 100124504-100124504 on Build GRCh38
rs201532149 Chr.7: 100124645-100124645 on Build GRCh38
rs764656853 Chr.7: 100124557-100124557 on Build GRCh38
rs757977425 Chr.7: 100124574-100124574 on Build GRCh38
rs776498769 Chr.7: 100124602-100124602 on Build GRCh38
rs750734379 Chr.7: 100124667-100124667 on Build GRCh38
rs761552985 Chr.7: 100124534-100124534 on Build GRCh38
rs769508965 Chr.7: 100124609-100124609 on Build GRCh38
rs376839210 Chr.7: 100124637-100124637 on Build GRCh38
rs754277562 Chr.7: 100124564-100124564 on Build GRCh38
rs759537243 Chr.7: 100124545-100124545 on Build GRCh38
rs892315588 Chr.7: 100124665-100124665 on Build GRCh38
rs758113251 Chr.7: 100124680-100124680 on Build GRCh38
rs777094819 Chr.7: 100124693-100124694 on Build GRCh38
rs1880949 Chr.7: 100124643-100124643 on Build GRCh38
rs771546278 Chr.7: 100124512-100124512 on Build GRCh38
rs760161046 Chr.7: 100124639-100124640 on Build GRCh38
rs770480313 Chr.7: 100124648-100124652 on Build GRCh38
rs199801822 Chr.7: 100124638-100124638 on Build GRCh38
rs757837953 Chr.7: 100124657-100124657 on Build GRCh38
rs776446653 Chr.7: 100124525-100124525 on Build GRCh38
rs779528954 Chr.7: 100124682-100124682 on Build GRCh38
rs369794793 Chr.7: 100124669-100124669 on Build GRCh38
rs754332452 Chr.7: 100124655-100124655 on Build GRCh38
rs377566765 Chr.7: 100124654-100124654 on Build GRCh38
rs774861124 Chr.7: 100124517-100124517 on Build GRCh38
rs1057024481 Chr.7: 100124583-100124583 on Build GRCh38
rs377668978 Chr.7: 100124567-100124567 on Build GRCh38
rs754424900 Chr.7: 100124699-100124699 on Build GRCh38
rs772803925 Chr.7: 100124530-100124533 on Build GRCh38
rs370237867 Chr.7: 100124698-100124698 on Build GRCh38
rs779406939 Chr.7: 100124706-100124706 on Build GRCh38
rs772985926 Chr.7: 100124630-100124630 on Build GRCh38
rs747947661 Chr.7: 100124608-100124608 on Build GRCh38
rs768607465 Chr.7: 100124588-100124588 on Build GRCh38
rs374822757 Chr.7: 100124565-100124565 on Build GRCh38
rs746474438 Chr.7: 100124581-100124581 on Build GRCh38
rs60551236 Chr.7: 100124550-100124550 on Build GRCh38
rs151184122 Chr.7: 100124570-100124570 on Build GRCh38
rs746547214 Chr.7: 100124693-100124693 on Build GRCh38
rs760726881 Chr.7: 100124521-100124521 on Build GRCh38
rs766607442 Chr.7: 100124536-100124536 on Build GRCh38
rs760069201 Chr.7: 100124708-100124709 on Build GRCh38
rs751721241 Chr.7: 100124544-100124544 on Build GRCh38
rs373590886 Chr.7: 100124498-100124498 on Build GRCh38
rs569434978 Chr.7: 100124575-100124575 on Build GRCh38
rs765758033 Chr.7: 100124662-100124662 on Build GRCh38
rs756693085 Chr.7: 100124554-100124554 on Build GRCh38
rs778662129 Chr.7: 100124502-100124502 on Build GRCh38
rs768459443 Chr.7: 100124524-100124524 on Build GRCh38

View Full Product Details