Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30927598-30927829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL19
Entrez Gene ID
54620
Gene Name
F-box and leucine rich repeat protein 19
Gene Aliases
CXXC11, Fbl19, JHDM1C
UniGene
Hs.152149
Gene Chromosome Location
Chr.16: 30923055-30948783 on Build GRCh38
Transcripts
NM_001099784.2 NM_001282351.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7323422 c.524G>C p.S175T Chr.16: 30927800-30927800 on Build GRCh38
6555632 c.447C>T p.T149T Chr.16: 30927638-30927638 on Build GRCh38
5510807 c.510C>T p.G170G Chr.16: 30927786-30927786 on Build GRCh38
5731322 c.473C>T p.S158L Chr.16: 30927749-30927749 on Build GRCh38
970021 c.518G>A p.G173D Chr.16: 30927794-30927794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781059280 Chr.16: 30927648-30927648 on Build GRCh38
rs374725615 Chr.16: 30927720-30927720 on Build GRCh38
rs765904924 Chr.16: 30927725-30927725 on Build GRCh38
rs777618399 Chr.16: 30927667-30927667 on Build GRCh38
rs1026282852 Chr.16: 30927653-30927653 on Build GRCh38
rs766585656 Chr.16: 30927749-30927749 on Build GRCh38
rs988896137 Chr.16: 30927807-30927807 on Build GRCh38
rs367877670 Chr.16: 30927777-30927777 on Build GRCh38
rs749016312 Chr.16: 30927722-30927722 on Build GRCh38
rs749574753 Chr.16: 30927802-30927802 on Build GRCh38
rs749093651 Chr.16: 30927673-30927673 on Build GRCh38
rs925706551 Chr.16: 30927800-30927800 on Build GRCh38
rs756087166 Chr.16: 30927652-30927652 on Build GRCh38
rs759805388 Chr.16: 30927771-30927771 on Build GRCh38
rs777252690 Chr.16: 30927664-30927668 on Build GRCh38
rs200017689 Chr.16: 30927641-30927641 on Build GRCh38
rs866602269 Chr.16: 30927619-30927619 on Build GRCh38
rs574467573 Chr.16: 30927699-30927699 on Build GRCh38
rs773572642 Chr.16: 30927729-30927729 on Build GRCh38
rs755082599 Chr.16: 30927647-30927647 on Build GRCh38
rs748057914 Chr.16: 30927650-30927650 on Build GRCh38
rs375367725 Chr.16: 30927722-30927722 on Build GRCh38
rs964502926 Chr.16: 30927766-30927766 on Build GRCh38
rs201806396 Chr.16: 30927684-30927684 on Build GRCh38
rs763236103 Chr.16: 30927748-30927748 on Build GRCh38
rs773876689 Chr.16: 30927675-30927675 on Build GRCh38
rs775186468 Chr.16: 30927713-30927713 on Build GRCh38
rs770558884 Chr.16: 30927674-30927674 on Build GRCh38
rs1027602681 Chr.16: 30927784-30927784 on Build GRCh38
rs751753002 Chr.16: 30927761-30927761 on Build GRCh38

View Full Product Details