Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 59464426-59464663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR135
Entrez Gene ID
64582
Gene Name
G protein-coupled receptor 135
Gene Aliases
HUMNPIIY20
UniGene
Hs.647573
Gene Chromosome Location
Chr.14: 59452773-59465356 on Build GRCh38
Transcripts
NM_022571.5 XM_017021596.1 XM_017021597.1 XM_017021598.1 XM_017021599.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6408906 c.692C>T p.A231V Chr.14: 59464535-59464535 on Build GRCh38
4989093 c.665G>A p.G222D Chr.14: 59464562-59464562 on Build GRCh38
399307 c.640G>A p.V214M Chr.14: 59464587-59464587 on Build GRCh38
6521537 c.644G>T p.R215L Chr.14: 59464583-59464583 on Build GRCh38
4577918 c.635C>T p.A212V Chr.14: 59464592-59464592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757418545 Chr.14: 59464448-59464448 on Build GRCh38
rs907616963 Chr.14: 59464485-59464485 on Build GRCh38
rs146353313 Chr.14: 59464631-59464631 on Build GRCh38
rs758385032 Chr.14: 59464567-59464567 on Build GRCh38
rs747448653 Chr.14: 59464523-59464523 on Build GRCh38
rs780380112 Chr.14: 59464580-59464580 on Build GRCh38
rs754412213 Chr.14: 59464561-59464561 on Build GRCh38
rs779079458 Chr.14: 59464462-59464462 on Build GRCh38
rs766875214 Chr.14: 59464605-59464605 on Build GRCh38
rs773646614 Chr.14: 59464445-59464445 on Build GRCh38
rs375838132 Chr.14: 59464563-59464563 on Build GRCh38
rs981796318 Chr.14: 59464530-59464530 on Build GRCh38
rs958700359 Chr.14: 59464634-59464634 on Build GRCh38
rs762342868 Chr.14: 59464529-59464529 on Build GRCh38
rs996251889 Chr.14: 59464521-59464521 on Build GRCh38
rs1040441008 Chr.14: 59464494-59464494 on Build GRCh38
rs780904264 Chr.14: 59464601-59464601 on Build GRCh38
rs1005254206 Chr.14: 59464463-59464463 on Build GRCh38
rs758569048 Chr.14: 59464643-59464643 on Build GRCh38
rs35719242 Chr.14: 59464593-59464594 on Build GRCh38
rs769218971 Chr.14: 59464554-59464555 on Build GRCh38
rs761669440 Chr.14: 59464501-59464501 on Build GRCh38
rs368276153 Chr.14: 59464584-59464584 on Build GRCh38
rs553482475 Chr.14: 59464610-59464610 on Build GRCh38
rs776615679 Chr.14: 59464499-59464499 on Build GRCh38
rs764737680 Chr.14: 59464510-59464510 on Build GRCh38
rs756571759 Chr.14: 59464554-59464554 on Build GRCh38
rs1054681594 Chr.14: 59464552-59464552 on Build GRCh38
rs1010393888 Chr.14: 59464564-59464564 on Build GRCh38
rs202025710 Chr.14: 59464562-59464562 on Build GRCh38
rs766109036 Chr.14: 59464540-59464540 on Build GRCh38
rs772298366 Chr.14: 59464468-59464468 on Build GRCh38
rs768499613 Chr.14: 59464587-59464587 on Build GRCh38
rs375205184 Chr.14: 59464619-59464619 on Build GRCh38
rs748101215 Chr.14: 59464602-59464602 on Build GRCh38
rs746870013 Chr.14: 59464497-59464497 on Build GRCh38
rs779930164 Chr.14: 59464644-59464644 on Build GRCh38
rs762587119 Chr.14: 59464607-59464607 on Build GRCh38
rs111917202 Chr.14: 59464609-59464609 on Build GRCh38
rs751161634 Chr.14: 59464549-59464549 on Build GRCh38
rs532792108 Chr.14: 59464486-59464486 on Build GRCh38
rs552448044 Chr.14: 59464500-59464500 on Build GRCh38
rs772656205 Chr.14: 59464518-59464518 on Build GRCh38

View Full Product Details