Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184317626-184317851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4G1
Entrez Gene ID
1981
Gene Name
eukaryotic translation initiation factor 4 gamma 1
Gene Aliases
EIF-4G1, EIF4F, EIF4G, EIF4GI, P220, PARK18
UniGene
Hs.433750
Gene Chromosome Location
Chr.3: 184314495-184335358 on Build GRCh38
Transcripts
NM_001194946.1 NM_001194947.1 NM_001291157.1 NM_182917.4 NM_198241.2 NM_198242.2 NM_198244.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5449831 c.362C>T p.P121L Chr.3: 184317754-184317754 on Build GRCh38
3331133 c.346C>T p.P116S Chr.3: 184317738-184317738 on Build GRCh38
4945310 c.381A>T p.P127P Chr.3: 184317773-184317773 on Build GRCh38
3331134 c.347C>T p.P116L Chr.3: 184317739-184317739 on Build GRCh38
4115737 c.329G>A p.R110H Chr.3: 184317721-184317721 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536804820 Chr.3: 184317663-184317663 on Build GRCh38
rs769269958 Chr.3: 184317783-184317783 on Build GRCh38
rs772383016 Chr.3: 184317699-184317700 on Build GRCh38
rs376912636 Chr.3: 184317764-184317764 on Build GRCh38
rs201488213 Chr.3: 184317688-184317688 on Build GRCh38
rs1014403591 Chr.3: 184317670-184317670 on Build GRCh38
rs761026770 Chr.3: 184317702-184317707 on Build GRCh38
rs1009464804 Chr.3: 184317791-184317791 on Build GRCh38
rs1021306052 Chr.3: 184317811-184317811 on Build GRCh38
rs757531274 Chr.3: 184317719-184317719 on Build GRCh38
rs983619664 Chr.3: 184317826-184317826 on Build GRCh38
rs866648432 Chr.3: 184317778-184317778 on Build GRCh38
rs761359067 Chr.3: 184317815-184317815 on Build GRCh38
rs748345765 Chr.3: 184317695-184317695 on Build GRCh38
rs775969277 Chr.3: 184317812-184317812 on Build GRCh38
rs372384604 Chr.3: 184317697-184317697 on Build GRCh38
rs775940503 Chr.3: 184317756-184317756 on Build GRCh38
rs754955324 Chr.3: 184317704-184317704 on Build GRCh38
rs777701425 Chr.3: 184317712-184317712 on Build GRCh38
rs59108039 Chr.3: 184317660-184317660 on Build GRCh38
rs746125036 Chr.3: 184317731-184317731 on Build GRCh38
rs905996095 Chr.3: 184317682-184317682 on Build GRCh38
rs775942553 Chr.3: 184317700-184317701 on Build GRCh38
rs369147609 Chr.3: 184317824-184317824 on Build GRCh38
rs755318821 Chr.3: 184317671-184317671 on Build GRCh38
rs994503850 Chr.3: 184317753-184317753 on Build GRCh38
rs772261137 Chr.3: 184317740-184317740 on Build GRCh38
rs762629982 Chr.3: 184317688-184317690 on Build GRCh38

View Full Product Details