Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55941488-55941719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL6ST
Entrez Gene ID
3572
Gene Name
interleukin 6 signal transducer
Gene Aliases
CD130, CDW130, GP130, IL-6RB
UniGene
Hs.532082
Gene Chromosome Location
Chr.5: 55935095-55994993 on Build GRCh38
Transcripts
NM_001190981.1 NM_002184.3 NM_175767.2 NR_120480.1 XM_017009441.1 XM_017009442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3617178 c.2122C>T p.L708L Chr.5: 55941717-55941717 on Build GRCh38
249426 c.2125A>T p.K709* Chr.5: 55941714-55941714 on Build GRCh38
3855685 c.2201T>C p.M734T Chr.5: 55941638-55941638 on Build GRCh38
6685443 c.2174G>T p.S725I Chr.5: 55941665-55941665 on Build GRCh38
5017798 c.2190delG p.S731fs*57 Chr.5: 55941649-55941649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886389172 Chr.5: 55941553-55941553 on Build GRCh38
rs756696579 Chr.5: 55941596-55941596 on Build GRCh38
rs751591161 Chr.5: 55941523-55941523 on Build GRCh38
rs749517791 Chr.5: 55941618-55941618 on Build GRCh38
rs773888810 Chr.5: 55941684-55941684 on Build GRCh38
rs146333104 Chr.5: 55941673-55941673 on Build GRCh38
rs1020528370 Chr.5: 55941676-55941676 on Build GRCh38
rs768990057 Chr.5: 55941628-55941628 on Build GRCh38
rs768129650 Chr.5: 55941524-55941524 on Build GRCh38
rs750944841 Chr.5: 55941525-55941525 on Build GRCh38
rs371804310 Chr.5: 55941635-55941635 on Build GRCh38
rs755954921 Chr.5: 55941677-55941677 on Build GRCh38
rs957509405 Chr.5: 55941643-55941643 on Build GRCh38
rs1001298767 Chr.5: 55941626-55941626 on Build GRCh38
rs140896990 Chr.5: 55941664-55941664 on Build GRCh38
rs866312303 Chr.5: 55941578-55941578 on Build GRCh38
rs745763248 Chr.5: 55941605-55941605 on Build GRCh38
rs138205220 Chr.5: 55941607-55941607 on Build GRCh38
rs375938437 Chr.5: 55941577-55941577 on Build GRCh38
rs61755738 Chr.5: 55941604-55941604 on Build GRCh38
rs774424754 Chr.5: 55941638-55941638 on Build GRCh38
rs752425339 Chr.5: 55941565-55941565 on Build GRCh38
rs746583730 Chr.5: 55941594-55941594 on Build GRCh38
rs756589757 Chr.5: 55941538-55941538 on Build GRCh38
rs866712918 Chr.5: 55941652-55941652 on Build GRCh38
rs758014866 Chr.5: 55941573-55941573 on Build GRCh38
rs780528560 Chr.5: 55941556-55941556 on Build GRCh38
rs764093325 Chr.5: 55941513-55941513 on Build GRCh38
rs61740791 Chr.5: 55941629-55941629 on Build GRCh38

View Full Product Details