Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1399109-1399338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAMT
Entrez Gene ID
2593
Gene Name
guanidinoacetate N-methyltransferase
Gene Aliases
CCDS2, HEL-S-20, PIG2, TP53I2
UniGene
Hs.81131
Gene Chromosome Location
Chr.19: 1397026-1401570 on Build GRCh38
Transcripts
NM_000156.5 NM_138924.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6575292 c.392-4G>A p.? Chr.19: 1399199-1399199 on Build GRCh38
6575293 c.392-4G>A p.? Chr.19: 1399199-1399199 on Build GRCh38
3719115 c.416delT p.L139fs*22 Chr.19: 1399171-1399171 on Build GRCh38
4746485 c.413delC p.P138fs*23 Chr.19: 1399174-1399174 on Build GRCh38
3692398 c.457A>G p.K153E Chr.19: 1399130-1399130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778443679 Chr.19: 1399312-1399312 on Build GRCh38
rs770240434 Chr.19: 1399300-1399300 on Build GRCh38
rs747656257 Chr.19: 1399168-1399168 on Build GRCh38
rs764210675 Chr.19: 1399200-1399200 on Build GRCh38
rs962865159 Chr.19: 1399193-1399193 on Build GRCh38
rs971660690 Chr.19: 1399273-1399273 on Build GRCh38
rs149821870 Chr.19: 1399150-1399150 on Build GRCh38
rs974222710 Chr.19: 1399316-1399316 on Build GRCh38
rs192416474 Chr.19: 1399202-1399202 on Build GRCh38
rs746251691 Chr.19: 1399159-1399159 on Build GRCh38
rs1031492095 Chr.19: 1399214-1399214 on Build GRCh38
rs200593372 Chr.19: 1399231-1399231 on Build GRCh38
rs374724533 Chr.19: 1399180-1399180 on Build GRCh38
rs199732257 Chr.19: 1399240-1399240 on Build GRCh38
rs773648628 Chr.19: 1399213-1399214 on Build GRCh38
rs1035762506 Chr.19: 1399290-1399290 on Build GRCh38
rs1002165125 Chr.19: 1399288-1399288 on Build GRCh38
rs373600604 Chr.19: 1399218-1399218 on Build GRCh38
rs920116948 Chr.19: 1399314-1399314 on Build GRCh38
rs536047178 Chr.19: 1399206-1399206 on Build GRCh38
rs1024750817 Chr.19: 1399272-1399272 on Build GRCh38
rs780692296 Chr.19: 1399219-1399219 on Build GRCh38
rs372673897 Chr.19: 1399161-1399161 on Build GRCh38
rs201528279 Chr.19: 1399138-1399138 on Build GRCh38
rs753979877 Chr.19: 1399201-1399201 on Build GRCh38
rs556829801 Chr.19: 1399185-1399185 on Build GRCh38
rs766918828 Chr.19: 1399239-1399240 on Build GRCh38
rs779376340 Chr.19: 1399158-1399158 on Build GRCh38
rs181832825 Chr.19: 1399315-1399315 on Build GRCh38
rs752148967 Chr.19: 1399240-1399249 on Build GRCh38
rs942634281 Chr.19: 1399220-1399220 on Build GRCh38
rs80338733 Chr.19: 1399149-1399149 on Build GRCh38
rs756178130 Chr.19: 1399195-1399195 on Build GRCh38
rs1034585807 Chr.19: 1399293-1399293 on Build GRCh38
rs774144200 Chr.19: 1399184-1399184 on Build GRCh38
rs761047151 Chr.19: 1399221-1399232 on Build GRCh38
rs1010235535 Chr.19: 1399270-1399270 on Build GRCh38
rs891893893 Chr.19: 1399259-1399259 on Build GRCh38
rs770557016 Chr.19: 1399239-1399239 on Build GRCh38
rs769395857 Chr.19: 1399230-1399230 on Build GRCh38
rs961686970 Chr.19: 1399294-1399294 on Build GRCh38
rs747568413 Chr.19: 1399221-1399221 on Build GRCh38
rs769485927 Chr.19: 1399171-1399171 on Build GRCh38
rs772974568 Chr.19: 1399179-1399179 on Build GRCh38
rs776155730 Chr.19: 1399167-1399167 on Build GRCh38
rs759367191 Chr.19: 1399245-1399245 on Build GRCh38
rs200500835 Chr.19: 1399191-1399191 on Build GRCh38
rs981094897 Chr.19: 1399284-1399284 on Build GRCh38
rs772373349 Chr.19: 1399143-1399145 on Build GRCh38

View Full Product Details