Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 74738277-74738548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFM2
Entrez Gene ID
84340
Gene Name
G elongation factor mitochondrial 2
Gene Aliases
EF-G2mt, EFG2, MRRF2, MST027, MSTP027, RRF2, RRF2mt, hEFG2, mEF-G 2
UniGene
Hs.277154
Gene Chromosome Location
Chr.5: 74721204-74767371 on Build GRCh38
Transcripts
NM_001281302.1 NM_032380.4 NM_170681.2 NM_170691.2 NR_104006.1 XM_011543691.2 XM_017009986.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5512116 c.1283C>T p.T428I Chr.5: 74738355-74738355 on Build GRCh38
5928680 c.1273C>T p.P425S Chr.5: 74738365-74738365 on Build GRCh38
5928679 c.1274C>T p.P425L Chr.5: 74738364-74738364 on Build GRCh38
6353673 c.1196T>C p.I399T Chr.5: 74738526-74738526 on Build GRCh38
5652030 c.1213A>C p.N405H Chr.5: 74738509-74738509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763038574 Chr.5: 74738523-74738523 on Build GRCh38
rs753884229 Chr.5: 74738390-74738390 on Build GRCh38
rs149163380 Chr.5: 74738365-74738365 on Build GRCh38
rs761036917 Chr.5: 74738380-74738380 on Build GRCh38
rs78674702 Chr.5: 74738462-74738462 on Build GRCh38
rs759625706 Chr.5: 74738502-74738502 on Build GRCh38
rs773425115 Chr.5: 74738492-74738492 on Build GRCh38
rs765235690 Chr.5: 74738507-74738507 on Build GRCh38
rs767704309 Chr.5: 74738374-74738374 on Build GRCh38
rs772506292 Chr.5: 74738489-74738489 on Build GRCh38
rs747760119 Chr.5: 74738486-74738486 on Build GRCh38
rs897046003 Chr.5: 74738412-74738412 on Build GRCh38
rs752948580 Chr.5: 74738514-74738514 on Build GRCh38
rs773797714 Chr.5: 74738375-74738375 on Build GRCh38
rs562244099 Chr.5: 74738315-74738315 on Build GRCh38
rs529758134 Chr.5: 74738330-74738330 on Build GRCh38
rs779755199 Chr.5: 74738457-74738457 on Build GRCh38
rs776748857 Chr.5: 74738501-74738501 on Build GRCh38
rs1017803669 Chr.5: 74738482-74738482 on Build GRCh38
rs182174189 Chr.5: 74738391-74738391 on Build GRCh38
rs375996126 Chr.5: 74738464-74738464 on Build GRCh38
rs868171310 Chr.5: 74738392-74738392 on Build GRCh38
rs765124597 Chr.5: 74738353-74738356 on Build GRCh38
rs773010798 Chr.5: 74738499-74738500 on Build GRCh38
rs887883100 Chr.5: 74738463-74738463 on Build GRCh38
rs772191981 Chr.5: 74738433-74738433 on Build GRCh38
rs371508939 Chr.5: 74738327-74738327 on Build GRCh38
rs201204413 Chr.5: 74738493-74738493 on Build GRCh38
rs142425196 Chr.5: 74738404-74738404 on Build GRCh38
rs1027641865 Chr.5: 74738427-74738427 on Build GRCh38
rs766791577 Chr.5: 74738495-74738495 on Build GRCh38
rs863224037 Chr.5: 74738409-74738409 on Build GRCh38
rs766526804 Chr.5: 74738382-74738382 on Build GRCh38
rs981113569 Chr.5: 74738448-74738448 on Build GRCh38
rs200232238 Chr.5: 74738413-74738413 on Build GRCh38
rs768396838 Chr.5: 74738340-74738340 on Build GRCh38
rs755985449 Chr.5: 74738442-74738442 on Build GRCh38
rs17852780 Chr.5: 74738364-74738364 on Build GRCh38
rs996253429 Chr.5: 74738354-74738354 on Build GRCh38
rs186213016 Chr.5: 74738465-74738465 on Build GRCh38
rs774926981 Chr.5: 74738360-74738360 on Build GRCh38
rs779489724 Chr.5: 74738319-74738319 on Build GRCh38
rs765432280 Chr.5: 74738398-74738398 on Build GRCh38
rs7732843 Chr.5: 74738420-74738420 on Build GRCh38
rs571673017 Chr.5: 74738403-74738403 on Build GRCh38
rs536132088 Chr.5: 74738430-74738430 on Build GRCh38

View Full Product Details