Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 211383458-211383719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERBB4
Entrez Gene ID
2066
Gene Name
erb-b2 receptor tyrosine kinase 4
Gene Aliases
ALS19, HER4, p180erbB4
UniGene
Hs.390729
Gene Chromosome Location
Chr.2: 211375717-212539147 on Build GRCh38
Transcripts
NM_001042599.1 NM_005235.2 XM_005246376.2 XM_005246377.2 XM_006712364.2 XM_017003577.1 XM_017003578.1 XM_017003579.1 XM_017003580.1 XM_017003581.1 XM_017003582.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6655841 c.3861G>C p.P1287P Chr.2: 211383651-211383651 on Build GRCh38
6587850 c.3847G>A p.E1283K Chr.2: 211383695-211383695 on Build GRCh38
2715852 c.3860C>T p.P1287L Chr.2: 211383652-211383652 on Build GRCh38
1205586 c.3902A>G p.Y1301C Chr.2: 211383640-211383640 on Build GRCh38
6156786 c.3879C>A p.H1293Q Chr.2: 211383633-211383633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778995420 Chr.2: 211383688-211383688 on Build GRCh38
rs878972139 Chr.2: 211383699-211383699 on Build GRCh38
rs758510094 Chr.2: 211383651-211383651 on Build GRCh38
rs752842064 Chr.2: 211383639-211383639 on Build GRCh38
rs745736412 Chr.2: 211383693-211383695 on Build GRCh38
rs749222561 Chr.2: 211383681-211383681 on Build GRCh38
rs751834116 Chr.2: 211383652-211383652 on Build GRCh38
rs1008338447 Chr.2: 211383557-211383558 on Build GRCh38
rs899718455 Chr.2: 211383581-211383581 on Build GRCh38
rs920954478 Chr.2: 211383529-211383529 on Build GRCh38
rs920182522 Chr.2: 211383507-211383508 on Build GRCh38
rs868458258 Chr.2: 211383695-211383695 on Build GRCh38
rs760087458 Chr.2: 211383631-211383631 on Build GRCh38
rs373629984 Chr.2: 211383601-211383601 on Build GRCh38
rs931272563 Chr.2: 211383518-211383518 on Build GRCh38
rs367568427 Chr.2: 211383683-211383683 on Build GRCh38
rs545302445 Chr.2: 211383664-211383664 on Build GRCh38
rs780037474 Chr.2: 211383675-211383675 on Build GRCh38
rs879163362 Chr.2: 211383632-211383632 on Build GRCh38
rs375899745 Chr.2: 211383633-211383633 on Build GRCh38
rs747831990 Chr.2: 211383569-211383569 on Build GRCh38
rs772930625 Chr.2: 211383604-211383604 on Build GRCh38
rs777088560 Chr.2: 211383620-211383620 on Build GRCh38
rs748475478 Chr.2: 211383692-211383692 on Build GRCh38
rs747549700 Chr.2: 211383690-211383690 on Build GRCh38
rs760895448 Chr.2: 211383608-211383608 on Build GRCh38
rs771634130 Chr.2: 211383698-211383698 on Build GRCh38
rs554806031 Chr.2: 211383556-211383556 on Build GRCh38
rs755829219 Chr.2: 211383654-211383654 on Build GRCh38
rs1045301588 Chr.2: 211383504-211383504 on Build GRCh38
rs766758455 Chr.2: 211383618-211383618 on Build GRCh38
rs149423647 Chr.2: 211383558-211383558 on Build GRCh38
rs376832876 Chr.2: 211383607-211383607 on Build GRCh38
rs955066264 Chr.2: 211383541-211383541 on Build GRCh38
rs1047074692 Chr.2: 211383523-211383523 on Build GRCh38
rs754085218 Chr.2: 211383557-211383557 on Build GRCh38

View Full Product Details