Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 151181315-151181542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR50
Entrez Gene ID
9248
Gene Name
G protein-coupled receptor 50
Gene Aliases
H9, Mel1c
UniGene
Hs.567390
Gene Chromosome Location
Chr.X: 151176584-151182855 on Build GRCh38
Transcripts
NM_004224.3 XM_011531216.2

SNP Detail

SNP ID Chromosome Location
rs894916748 Chr.X: 151181472-151181472 on Build GRCh38
rs749220529 Chr.X: 151181452-151181452 on Build GRCh38
rs756799778 Chr.X: 151181437-151181437 on Build GRCh38
rs13440581 Chr.X: 151181399-151181399 on Build GRCh38
rs776869695 Chr.X: 151181359-151181359 on Build GRCh38
rs750125551 Chr.X: 151181446-151181446 on Build GRCh38
rs759844066 Chr.X: 151181392-151181392 on Build GRCh38
rs999863231 Chr.X: 151181471-151181471 on Build GRCh38
rs369231570 Chr.X: 151181504-151181504 on Build GRCh38
rs866973433 Chr.X: 151181386-151181386 on Build GRCh38
rs760795393 Chr.X: 151181349-151181353 on Build GRCh38
rs760818840 Chr.X: 151181336-151181336 on Build GRCh38
rs780058140 Chr.X: 151181449-151181449 on Build GRCh38
rs763439576 Chr.X: 151181412-151181412 on Build GRCh38
rs778913282 Chr.X: 151181466-151181466 on Build GRCh38
rs750977314 Chr.X: 151181432-151181432 on Build GRCh38
rs201645456 Chr.X: 151181393-151181393 on Build GRCh38
rs747020113 Chr.X: 151181469-151181469 on Build GRCh38
rs773218380 Chr.X: 151181335-151181335 on Build GRCh38
rs754810429 Chr.X: 151181459-151181459 on Build GRCh38
rs770991631 Chr.X: 151181351-151181351 on Build GRCh38
rs763647516 Chr.X: 151181417-151181417 on Build GRCh38
rs765410234 Chr.X: 151181345-151181345 on Build GRCh38
rs755938733 Chr.X: 151181448-151181448 on Build GRCh38
rs775560713 Chr.X: 151181493-151181493 on Build GRCh38
rs767243441 Chr.X: 151181444-151181444 on Build GRCh38

View Full Product Details