Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31392192-31392425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSG1-AS1
Entrez Gene ID
101927718
Gene Name
DSG1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.625473
Gene Chromosome Location
Chr.18: 31343368-31426988 on Build GRCh38
Transcripts
NR_110788.1 NM_001134453.2 NM_177986.4

Gene Symbol
DSG4
Entrez Gene ID
147409
Gene Name
desmoglein 4
Gene Aliases
CDGF13, CDHF13, HYPT6, LAH
UniGene
Hs.407618
Gene Chromosome Location
Chr.18: 31376777-31415791 on Build GRCh38
Transcripts
NR_110788.1 NM_001134453.2 NM_177986.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1290809 c.950G>T p.W317L Chr.18: 31392285-31392285 on Build GRCh38
3525114 c.944G>A p.G315E Chr.18: 31392279-31392279 on Build GRCh38
5681721 c.913G>T p.A305S Chr.18: 31392248-31392248 on Build GRCh38
5387978 c.935G>A p.G312E Chr.18: 31392270-31392270 on Build GRCh38
3525116 c.954C>T p.F318F Chr.18: 31392289-31392289 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759072801 Chr.18: 31392270-31392270 on Build GRCh38
rs767724665 Chr.18: 31392303-31392303 on Build GRCh38
rs757685916 Chr.18: 31392291-31392291 on Build GRCh38
rs781514202 Chr.18: 31392224-31392224 on Build GRCh38
rs764811428 Chr.18: 31392273-31392273 on Build GRCh38
rs749446111 Chr.18: 31392320-31392320 on Build GRCh38
rs149724713 Chr.18: 31392315-31392315 on Build GRCh38
rs146457133 Chr.18: 31392265-31392265 on Build GRCh38
rs750686825 Chr.18: 31392305-31392305 on Build GRCh38
rs774927290 Chr.18: 31392275-31392275 on Build GRCh38
rs776227245 Chr.18: 31392372-31392372 on Build GRCh38
rs1039979540 Chr.18: 31392393-31392393 on Build GRCh38
rs764648086 Chr.18: 31392289-31392289 on Build GRCh38
rs886053705 Chr.18: 31392263-31392263 on Build GRCh38
rs780240568 Chr.18: 31392237-31392237 on Build GRCh38
rs748100051 Chr.18: 31392241-31392241 on Build GRCh38
rs770793846 Chr.18: 31392354-31392354 on Build GRCh38
rs745551490 Chr.18: 31392389-31392389 on Build GRCh38
rs550801669 Chr.18: 31392236-31392236 on Build GRCh38
rs756223021 Chr.18: 31392313-31392313 on Build GRCh38
rs774592364 Chr.18: 31392341-31392341 on Build GRCh38
rs773279288 Chr.18: 31392244-31392244 on Build GRCh38
rs941593482 Chr.18: 31392347-31392347 on Build GRCh38
rs139090943 Chr.18: 31392232-31392232 on Build GRCh38
rs746863735 Chr.18: 31392254-31392254 on Build GRCh38
rs908911212 Chr.18: 31392317-31392317 on Build GRCh38
rs770611447 Chr.18: 31392255-31392255 on Build GRCh38
rs375185491 Chr.18: 31392326-31392326 on Build GRCh38
rs17802140 Chr.18: 31392400-31392400 on Build GRCh38
rs772194792 Chr.18: 31392242-31392242 on Build GRCh38
rs989929740 Chr.18: 31392227-31392227 on Build GRCh38
rs755077799 Chr.18: 31392321-31392321 on Build GRCh38
rs267605157 Chr.18: 31392302-31392302 on Build GRCh38
rs762432591 Chr.18: 31392277-31392277 on Build GRCh38
rs140750904 Chr.18: 31392290-31392290 on Build GRCh38

View Full Product Details