Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30006220-30006475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOC2A
Entrez Gene ID
8448
Gene Name
double C2 domain alpha
Gene Aliases
Doc2
UniGene
Hs.355281
Gene Chromosome Location
Chr.16: 30005514-30023280 on Build GRCh38
Transcripts
NM_001282062.1 NM_001282063.1 NM_001282068.1 NM_003586.2 NR_104089.1 NR_104090.1 XM_011545975.1 XM_017023776.1 XM_017023777.1 XM_017023778.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7286489 c.1087G>T p.G363C Chr.16: 30006302-30006302 on Build GRCh38
1377315 c.1097G>A p.R366Q Chr.16: 30006292-30006292 on Build GRCh38
5462526 c.1015A>G p.T339A Chr.16: 30006455-30006455 on Build GRCh38
4390062 c.1097G>T p.R366L Chr.16: 30006292-30006292 on Build GRCh38
292141 c.1044C>A p.S348S Chr.16: 30006426-30006426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774531629 Chr.16: 30006319-30006319 on Build GRCh38
rs1020441474 Chr.16: 30006302-30006302 on Build GRCh38
rs771408208 Chr.16: 30006314-30006314 on Build GRCh38
rs145044673 Chr.16: 30006304-30006304 on Build GRCh38
rs779240266 Chr.16: 30006394-30006394 on Build GRCh38
rs1013134431 Chr.16: 30006393-30006393 on Build GRCh38
rs369114686 Chr.16: 30006264-30006264 on Build GRCh38
rs761033571 Chr.16: 30006436-30006436 on Build GRCh38
rs779765863 Chr.16: 30006376-30006376 on Build GRCh38
rs200732948 Chr.16: 30006427-30006427 on Build GRCh38
rs764047517 Chr.16: 30006346-30006346 on Build GRCh38
rs1032013453 Chr.16: 30006280-30006280 on Build GRCh38
rs370177489 Chr.16: 30006326-30006326 on Build GRCh38
rs200262965 Chr.16: 30006245-30006245 on Build GRCh38
rs757306872 Chr.16: 30006350-30006350 on Build GRCh38
rs147113011 Chr.16: 30006263-30006263 on Build GRCh38
rs767804083 Chr.16: 30006327-30006327 on Build GRCh38
rs141936366 Chr.16: 30006392-30006392 on Build GRCh38
rs373250621 Chr.16: 30006292-30006292 on Build GRCh38
rs774283753 Chr.16: 30006260-30006260 on Build GRCh38
rs749624024 Chr.16: 30006391-30006391 on Build GRCh38
rs765050481 Chr.16: 30006353-30006353 on Build GRCh38
rs770312614 Chr.16: 30006248-30006248 on Build GRCh38
rs775517694 Chr.16: 30006340-30006340 on Build GRCh38
rs753671040 Chr.16: 30006349-30006349 on Build GRCh38
rs763028825 Chr.16: 30006257-30006257 on Build GRCh38
rs779529166 Chr.16: 30006298-30006298 on Build GRCh38
rs141323522 Chr.16: 30006299-30006299 on Build GRCh38
rs771978913 Chr.16: 30006411-30006411 on Build GRCh38
rs749855078 Chr.16: 30006307-30006307 on Build GRCh38
rs745974183 Chr.16: 30006403-30006403 on Build GRCh38
rs372707859 Chr.16: 30006244-30006244 on Build GRCh38
rs762033437 Chr.16: 30006361-30006362 on Build GRCh38
rs764139821 Chr.16: 30006267-30006267 on Build GRCh38
rs866898989 Chr.16: 30006419-30006419 on Build GRCh38
rs200796383 Chr.16: 30006452-30006452 on Build GRCh38
rs753760872 Chr.16: 30006277-30006277 on Build GRCh38
rs778162140 Chr.16: 30006383-30006383 on Build GRCh38
rs910381791 Chr.16: 30006297-30006297 on Build GRCh38
rs773655247 Chr.16: 30006256-30006256 on Build GRCh38
rs758106842 Chr.16: 30006295-30006295 on Build GRCh38
rs766515431 Chr.16: 30006258-30006258 on Build GRCh38
rs371322738 Chr.16: 30006261-30006261 on Build GRCh38
rs896078007 Chr.16: 30006387-30006387 on Build GRCh38
rs745345298 Chr.16: 30006293-30006293 on Build GRCh38
rs757255333 Chr.16: 30006284-30006284 on Build GRCh38
rs201063562 Chr.16: 30006367-30006367 on Build GRCh38
rs868040487 Chr.16: 30006310-30006310 on Build GRCh38
rs375392348 Chr.16: 30006359-30006359 on Build GRCh38
rs368650314 Chr.16: 30006379-30006379 on Build GRCh38
rs765248168 Chr.16: 30006376-30006379 on Build GRCh38
rs112422001 Chr.16: 30006342-30006342 on Build GRCh38
rs776765147 Chr.16: 30006305-30006306 on Build GRCh38
rs138847296 Chr.16: 30006453-30006453 on Build GRCh38
rs34246709 Chr.16: 30006262-30006262 on Build GRCh38
rs770081731 Chr.16: 30006300-30006300 on Build GRCh38
rs934666271 Chr.16: 30006358-30006358 on Build GRCh38
rs754615522 Chr.16: 30006380-30006380 on Build GRCh38

View Full Product Details