Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17338394-17338657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTPBP3
Entrez Gene ID
84705
Gene Name
GTP binding protein 3 (mitochondrial)
Gene Aliases
COXPD23, GTPBG3, MSS1, MTGP1, THDF1
UniGene
Hs.334885
Gene Chromosome Location
Chr.19: 17334982-17342731 on Build GRCh38
Transcripts
NM_001128855.2 NM_001195422.1 NM_032620.3 NM_133644.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5972772 c.423G>A p.P141P Chr.19: 17338420-17338420 on Build GRCh38
3718145 c.464T>A p.L155Q Chr.19: 17338614-17338614 on Build GRCh38
4689235 c.507G>A p.A169A Chr.19: 17338591-17338591 on Build GRCh38
7155102 c.444C>T p.F148F Chr.19: 17338594-17338594 on Build GRCh38
5940860 c.455-7C>T p.? Chr.19: 17338532-17338532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769418711 Chr.19: 17338572-17338572 on Build GRCh38
rs757131954 Chr.19: 17338635-17338635 on Build GRCh38
rs375701585 Chr.19: 17338551-17338551 on Build GRCh38
rs752018459 Chr.19: 17338527-17338527 on Build GRCh38
rs780563817 Chr.19: 17338564-17338564 on Build GRCh38
rs190556849 Chr.19: 17338514-17338514 on Build GRCh38
rs759044252 Chr.19: 17338499-17338499 on Build GRCh38
rs756369422 Chr.19: 17338432-17338432 on Build GRCh38
rs999488371 Chr.19: 17338552-17338552 on Build GRCh38
rs142059730 Chr.19: 17338570-17338570 on Build GRCh38
rs143759529 Chr.19: 17338419-17338419 on Build GRCh38
rs777733575 Chr.19: 17338559-17338559 on Build GRCh38
rs750455793 Chr.19: 17338608-17338608 on Build GRCh38
rs760855666 Chr.19: 17338613-17338613 on Build GRCh38
rs775823244 Chr.19: 17338505-17338505 on Build GRCh38
rs923024478 Chr.19: 17338414-17338414 on Build GRCh38
rs781353045 Chr.19: 17338517-17338517 on Build GRCh38
rs1054884170 Chr.19: 17338468-17338468 on Build GRCh38
rs370787785 Chr.19: 17338475-17338475 on Build GRCh38
rs774708853 Chr.19: 17338590-17338590 on Build GRCh38
rs146343760 Chr.19: 17338634-17338634 on Build GRCh38
rs549090990 Chr.19: 17338433-17338433 on Build GRCh38
rs372721127 Chr.19: 17338618-17338618 on Build GRCh38
rs372521902 Chr.19: 17338490-17338490 on Build GRCh38
rs730880255 Chr.19: 17338626-17338626 on Build GRCh38
rs376384130 Chr.19: 17338557-17338557 on Build GRCh38
rs770300748 Chr.19: 17338489-17338489 on Build GRCh38
rs762155226 Chr.19: 17338591-17338591 on Build GRCh38
rs748022451 Chr.19: 17338456-17338456 on Build GRCh38
rs1047386127 Chr.19: 17338436-17338436 on Build GRCh38
rs201509276 Chr.19: 17338523-17338524 on Build GRCh38
rs770254800 Chr.19: 17338431-17338433 on Build GRCh38
rs145625259 Chr.19: 17338579-17338579 on Build GRCh38
rs769560402 Chr.19: 17338501-17338501 on Build GRCh38
rs767434617 Chr.19: 17338542-17338542 on Build GRCh38
rs909096383 Chr.19: 17338462-17338462 on Build GRCh38
rs956975931 Chr.19: 17338612-17338612 on Build GRCh38
rs558202040 Chr.19: 17338614-17338614 on Build GRCh38
rs762403119 Chr.19: 17338532-17338532 on Build GRCh38
rs772120855 Chr.19: 17338464-17338464 on Build GRCh38
rs940351020 Chr.19: 17338437-17338437 on Build GRCh38
rs746975856 Chr.19: 17338484-17338484 on Build GRCh38
rs754066658 Chr.19: 17338615-17338615 on Build GRCh38
rs776029170 Chr.19: 17338489-17338490 on Build GRCh38
rs756284329 Chr.19: 17338548-17338548 on Build GRCh38
rs750517470 Chr.19: 17338544-17338544 on Build GRCh38
rs1032351386 Chr.19: 17338553-17338553 on Build GRCh38
rs764556394 Chr.19: 17338526-17338526 on Build GRCh38
rs770906277 Chr.19: 17338563-17338563 on Build GRCh38
rs376592241 Chr.19: 17338429-17338429 on Build GRCh38
rs777905189 Chr.19: 17338465-17338465 on Build GRCh38
rs781243432 Chr.19: 17338636-17338636 on Build GRCh38
rs764647980 Chr.19: 17338412-17338412 on Build GRCh38
rs772716704 Chr.19: 17338550-17338551 on Build GRCh38
rs745687275 Chr.19: 17338594-17338594 on Build GRCh38
rs746865025 Chr.19: 17338560-17338560 on Build GRCh38
rs746428014 Chr.19: 17338417-17338417 on Build GRCh38
rs768135810 Chr.19: 17338592-17338592 on Build GRCh38
rs777645307 Chr.19: 17338622-17338622 on Build GRCh38
rs762627810 Chr.19: 17338586-17338586 on Build GRCh38
rs751619113 Chr.19: 17338418-17338418 on Build GRCh38
rs768956976 Chr.19: 17338587-17338587 on Build GRCh38
rs1018844845 Chr.19: 17338620-17338620 on Build GRCh38
rs571877236 Chr.19: 17338522-17338523 on Build GRCh38

View Full Product Details