Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8003731-8003988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUCY2D
Entrez Gene ID
3000
Gene Name
guanylate cyclase 2D, retinal
Gene Aliases
CORD5, CORD6, CYGD, GUC1A4, GUC2D, LCA, LCA1, RCD2, RETGC-1, ROS-GC1, ROSGC, retGC
UniGene
Hs.592109
Gene Chromosome Location
Chr.17: 8002670-8020340 on Build GRCh38
Transcripts
NM_000180.3 XM_011523816.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2804360 c.759C>T p.G253G Chr.17: 8003889-8003889 on Build GRCh38
2804361 c.777C>T p.L259L Chr.17: 8003907-8003907 on Build GRCh38
3523474 c.815C>T p.S272F Chr.17: 8003945-8003945 on Build GRCh38
4070557 c.799G>A p.G267S Chr.17: 8003929-8003929 on Build GRCh38
5525033 c.686C>T p.A229V Chr.17: 8003733-8003733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755329222 Chr.17: 8003777-8003777 on Build GRCh38
rs890286878 Chr.17: 8003819-8003819 on Build GRCh38
rs766859707 Chr.17: 8003759-8003759 on Build GRCh38
rs141956583 Chr.17: 8003860-8003860 on Build GRCh38
rs768206746 Chr.17: 8003870-8003870 on Build GRCh38
rs746769693 Chr.17: 8003934-8003934 on Build GRCh38
rs755787827 Chr.17: 8003801-8003801 on Build GRCh38
rs770035607 Chr.17: 8003848-8003848 on Build GRCh38
rs886038268 Chr.17: 8003856-8003856 on Build GRCh38
rs138922415 Chr.17: 8003873-8003873 on Build GRCh38
rs760107054 Chr.17: 8003864-8003864 on Build GRCh38
rs148987106 Chr.17: 8003877-8003877 on Build GRCh38
rs775258389 Chr.17: 8003829-8003829 on Build GRCh38
rs773415492 Chr.17: 8003937-8003937 on Build GRCh38
rs3829789 Chr.17: 8003871-8003871 on Build GRCh38
rs762006599 Chr.17: 8003754-8003754 on Build GRCh38
rs773327031 Chr.17: 8003851-8003851 on Build GRCh38
rs776773228 Chr.17: 8003748-8003748 on Build GRCh38
rs763099634 Chr.17: 8003938-8003938 on Build GRCh38
rs759731211 Chr.17: 8003961-8003961 on Build GRCh38
rs753468278 Chr.17: 8003881-8003881 on Build GRCh38
rs778378811 Chr.17: 8003907-8003907 on Build GRCh38
rs750484873 Chr.17: 8003758-8003758 on Build GRCh38
rs745816355 Chr.17: 8003823-8003823 on Build GRCh38
rs771338362 Chr.17: 8003946-8003946 on Build GRCh38
rs768365237 Chr.17: 8003935-8003935 on Build GRCh38
rs61749672 Chr.17: 8003909-8003909 on Build GRCh38
rs763047690 Chr.17: 8003858-8003858 on Build GRCh38
rs760426420 Chr.17: 8003840-8003840 on Build GRCh38
rs778752848 Chr.17: 8003822-8003822 on Build GRCh38
rs774569101 Chr.17: 8003959-8003959 on Build GRCh38
rs899659483 Chr.17: 8003755-8003755 on Build GRCh38
rs772051916 Chr.17: 8003825-8003825 on Build GRCh38
rs770374274 Chr.17: 8003811-8003811 on Build GRCh38
rs557241953 Chr.17: 8003806-8003806 on Build GRCh38
rs959135126 Chr.17: 8003866-8003866 on Build GRCh38
rs777336060 Chr.17: 8003805-8003805 on Build GRCh38
rs1020620560 Chr.17: 8003809-8003809 on Build GRCh38
rs890858732 Chr.17: 8003795-8003795 on Build GRCh38
rs780014587 Chr.17: 8003932-8003932 on Build GRCh38
rs752057528 Chr.17: 8003862-8003862 on Build GRCh38
rs752141508 Chr.17: 8003768-8003768 on Build GRCh38
rs777436847 Chr.17: 8003880-8003880 on Build GRCh38
rs765439946 Chr.17: 8003757-8003757 on Build GRCh38
rs925889678 Chr.17: 8003786-8003786 on Build GRCh38
rs978420396 Chr.17: 8003785-8003785 on Build GRCh38
rs756730335 Chr.17: 8003890-8003890 on Build GRCh38
rs745444949 Chr.17: 8003917-8003917 on Build GRCh38
rs375760706 Chr.17: 8003798-8003798 on Build GRCh38
rs771962267 Chr.17: 8003926-8003926 on Build GRCh38
rs903470945 Chr.17: 8003818-8003818 on Build GRCh38
rs774434849 Chr.17: 8003809-8003837 on Build GRCh38
rs868557040 Chr.17: 8003969-8003969 on Build GRCh38
rs1027644020 Chr.17: 8003872-8003872 on Build GRCh38

View Full Product Details