Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157032743-157033016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAVCR1
Entrez Gene ID
26762
Gene Name
hepatitis A virus cellular receptor 1
Gene Aliases
CD365, HAVCR, HAVCR-1, KIM-1, KIM1, TIM, TIM-1, TIM1, TIMD-1, TIMD1
UniGene
Hs.129711
Gene Chromosome Location
Chr.5: 156979443-157059119 on Build GRCh38
Transcripts
NM_001173393.2 NM_001308156.1 NM_012206.3 XM_011534515.2 XM_017009339.1 XM_017009340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6348744 c.986+6C>A p.? Chr.5: 157032848-157032848 on Build GRCh38
4905226 c.969G>A p.K323K Chr.5: 157032871-157032871 on Build GRCh38
4003419 c.986+9G>A p.? Chr.5: 157032845-157032845 on Build GRCh38
5625310 c.963C>T p.F321F Chr.5: 157032877-157032877 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750011065 Chr.5: 157032811-157032811 on Build GRCh38
rs960448564 Chr.5: 157032812-157032812 on Build GRCh38
rs757686314 Chr.5: 157032859-157032859 on Build GRCh38
rs899259103 Chr.5: 157032861-157032861 on Build GRCh38
rs1016872704 Chr.5: 157032794-157032794 on Build GRCh38
rs369560465 Chr.5: 157032893-157032893 on Build GRCh38
rs770047531 Chr.5: 157032846-157032846 on Build GRCh38
rs776248942 Chr.5: 157032927-157032929 on Build GRCh38
rs776539060 Chr.5: 157032902-157032902 on Build GRCh38
rs563153494 Chr.5: 157032763-157032763 on Build GRCh38
rs761687646 Chr.5: 157032913-157032913 on Build GRCh38
rs980078986 Chr.5: 157032911-157032911 on Build GRCh38
rs185021127 Chr.5: 157032894-157032894 on Build GRCh38
rs551458871 Chr.5: 157032961-157032961 on Build GRCh38
rs375307674 Chr.5: 157032875-157032875 on Build GRCh38
rs1005902946 Chr.5: 157032787-157032787 on Build GRCh38
rs34670839 Chr.5: 157032845-157032845 on Build GRCh38
rs74607260 Chr.5: 157032802-157032802 on Build GRCh38
rs1053566309 Chr.5: 157032777-157032777 on Build GRCh38
rs368799243 Chr.5: 157032809-157032809 on Build GRCh38
rs757977179 Chr.5: 157032815-157032815 on Build GRCh38
rs193271240 Chr.5: 157032778-157032778 on Build GRCh38
rs760699658 Chr.5: 157032877-157032877 on Build GRCh38
rs1040354476 Chr.5: 157032964-157032966 on Build GRCh38
rs35340566 Chr.5: 157032961-157032961 on Build GRCh38
rs781641544 Chr.5: 157032837-157032837 on Build GRCh38
rs779638245 Chr.5: 157032818-157032818 on Build GRCh38
rs954714658 Chr.5: 157032956-157032956 on Build GRCh38
rs924250399 Chr.5: 157032951-157032951 on Build GRCh38
rs947744369 Chr.5: 157032790-157032790 on Build GRCh38
rs571436305 Chr.5: 157032969-157032969 on Build GRCh38
rs749450406 Chr.5: 157032868-157032868 on Build GRCh38
rs199541418 Chr.5: 157032823-157032823 on Build GRCh38
rs767892992 Chr.5: 157032861-157032861 on Build GRCh38
rs935325113 Chr.5: 157032954-157032955 on Build GRCh38
rs777971381 Chr.5: 157032857-157032857 on Build GRCh38
rs754558797 Chr.5: 157032801-157032801 on Build GRCh38
rs747562961 Chr.5: 157032822-157032822 on Build GRCh38
rs878965332 Chr.5: 157032906-157032906 on Build GRCh38

View Full Product Details