Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 121623119-121623380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO40
Entrez Gene ID
51725
Gene Name
F-box protein 40
Gene Aliases
FBX40
UniGene
Hs.272564
Gene Chromosome Location
Chr.3: 121593323-121630292 on Build GRCh38
Transcripts
NM_016298.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3585988 c.1874G>A p.R625K Chr.3: 121623303-121623303 on Build GRCh38
3585987 c.1863G>A p.W621* Chr.3: 121623292-121623292 on Build GRCh38
6836149 c.1724G>C p.S575T Chr.3: 121623153-121623153 on Build GRCh38
3585984 c.1693C>T p.H565Y Chr.3: 121623122-121623122 on Build GRCh38
4683997 c.1701delA p.S569fs*5 Chr.3: 121623130-121623130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776521736 Chr.3: 121623178-121623178 on Build GRCh38
rs769567262 Chr.3: 121623182-121623182 on Build GRCh38
rs746991047 Chr.3: 121623162-121623162 on Build GRCh38
rs756752342 Chr.3: 121623262-121623262 on Build GRCh38
rs147922715 Chr.3: 121623321-121623321 on Build GRCh38
rs751068036 Chr.3: 121623259-121623259 on Build GRCh38
rs142139104 Chr.3: 121623144-121623144 on Build GRCh38
rs34348750 Chr.3: 121623210-121623210 on Build GRCh38
rs184673980 Chr.3: 121623204-121623204 on Build GRCh38
rs773454337 Chr.3: 121623305-121623305 on Build GRCh38
rs757931041 Chr.3: 121623146-121623146 on Build GRCh38
rs145018488 Chr.3: 121623209-121623209 on Build GRCh38
rs781160734 Chr.3: 121623263-121623263 on Build GRCh38
rs35106461 Chr.3: 121623153-121623153 on Build GRCh38
rs973104741 Chr.3: 121623320-121623320 on Build GRCh38
rs745742906 Chr.3: 121623265-121623265 on Build GRCh38
rs770824338 Chr.3: 121623167-121623167 on Build GRCh38
rs200687176 Chr.3: 121623294-121623294 on Build GRCh38
rs748947214 Chr.3: 121623264-121623265 on Build GRCh38
rs528828647 Chr.3: 121623292-121623292 on Build GRCh38
rs765631213 Chr.3: 121623217-121623217 on Build GRCh38
rs973144231 Chr.3: 121623234-121623234 on Build GRCh38
rs1031021764 Chr.3: 121623158-121623158 on Build GRCh38
rs985073438 Chr.3: 121623322-121623322 on Build GRCh38
rs748427565 Chr.3: 121623278-121623278 on Build GRCh38
rs140154336 Chr.3: 121623186-121623186 on Build GRCh38
rs201570535 Chr.3: 121623154-121623154 on Build GRCh38
rs777458705 Chr.3: 121623246-121623246 on Build GRCh38
rs753051384 Chr.3: 121623341-121623341 on Build GRCh38
rs760734100 Chr.3: 121623307-121623307 on Build GRCh38
rs763343276 Chr.3: 121623343-121623343 on Build GRCh38
rs773119943 Chr.3: 121623200-121623200 on Build GRCh38
rs752424663 Chr.3: 121623230-121623230 on Build GRCh38
rs779097611 Chr.3: 121623179-121623179 on Build GRCh38
rs372813938 Chr.3: 121623211-121623211 on Build GRCh38
rs773570752 Chr.3: 121623179-121623179 on Build GRCh38
rs777291696 Chr.3: 121623318-121623318 on Build GRCh38
rs779787731 Chr.3: 121623277-121623277 on Build GRCh38
rs763718593 Chr.3: 121623347-121623347 on Build GRCh38
rs995796984 Chr.3: 121623280-121623280 on Build GRCh38
rs771121662 Chr.3: 121623316-121623316 on Build GRCh38
rs937419148 Chr.3: 121623163-121623163 on Build GRCh38
rs775756963 Chr.3: 121623331-121623331 on Build GRCh38
rs368217296 Chr.3: 121623199-121623199 on Build GRCh38
rs141873915 Chr.3: 121623330-121623330 on Build GRCh38
rs770641563 Chr.3: 121623338-121623339 on Build GRCh38
rs560066260 Chr.3: 121623238-121623238 on Build GRCh38
rs769585447 Chr.3: 121623270-121623270 on Build GRCh38
rs151167240 Chr.3: 121623160-121623160 on Build GRCh38

View Full Product Details