Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74866337-74866608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDXR
Entrez Gene ID
2232
Gene Name
ferredoxin reductase
Gene Aliases
ADXR
UniGene
Hs.69745
Gene Chromosome Location
Chr.17: 74862497-74873031 on Build GRCh38
Transcripts
NM_001258012.3 NM_001258013.3 NM_001258014.3 NM_001258015.2 NM_001258016.3 NM_004110.5 NM_024417.4 NR_047576.2 XM_006721772.1 XM_011524527.1 XM_011524528.1 XM_011524529.1 XM_011524531.2 XM_011524532.2 XM_011524533.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6147979 c.360G>T p.P120P Chr.17: 74866479-74866479 on Build GRCh38
1680076 c.311G>A p.R104H Chr.17: 74866528-74866528 on Build GRCh38
4271042 c.179G>A p.R60H Chr.17: 74866528-74866528 on Build GRCh38
4271043 c.164C>T p.T55M Chr.17: 74866543-74866543 on Build GRCh38
6401395 c.382G>T p.A128S Chr.17: 74866457-74866457 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7223627 Chr.17: 74866491-74866491 on Build GRCh38
rs551451161 Chr.17: 74866485-74866485 on Build GRCh38
rs761558804 Chr.17: 74866409-74866409 on Build GRCh38
rs778001476 Chr.17: 74866554-74866555 on Build GRCh38
rs146885267 Chr.17: 74866479-74866479 on Build GRCh38
rs766169513 Chr.17: 74866431-74866431 on Build GRCh38
rs918940252 Chr.17: 74866402-74866402 on Build GRCh38
rs690514 Chr.17: 74866471-74866471 on Build GRCh38
rs776714079 Chr.17: 74866529-74866529 on Build GRCh38
rs984749438 Chr.17: 74866457-74866457 on Build GRCh38
rs374317439 Chr.17: 74866458-74866458 on Build GRCh38
rs776221918 Chr.17: 74866398-74866398 on Build GRCh38
rs141174234 Chr.17: 74866543-74866543 on Build GRCh38
rs376477275 Chr.17: 74866435-74866435 on Build GRCh38
rs537929036 Chr.17: 74866472-74866472 on Build GRCh38
rs550083102 Chr.17: 74866464-74866464 on Build GRCh38
rs770691402 Chr.17: 74866514-74866514 on Build GRCh38
rs767404506 Chr.17: 74866528-74866528 on Build GRCh38
rs769402154 Chr.17: 74866490-74866490 on Build GRCh38
rs766656876 Chr.17: 74866580-74866580 on Build GRCh38
rs757503961 Chr.17: 74866460-74866460 on Build GRCh38
rs923683362 Chr.17: 74866426-74866426 on Build GRCh38
rs368300344 Chr.17: 74866385-74866385 on Build GRCh38
rs183154538 Chr.17: 74866515-74866515 on Build GRCh38
rs2070918 Chr.17: 74866583-74866583 on Build GRCh38
rs764175967 Chr.17: 74866456-74866456 on Build GRCh38
rs1033305060 Chr.17: 74866548-74866548 on Build GRCh38
rs965793415 Chr.17: 74866386-74866386 on Build GRCh38
rs765295168 Chr.17: 74866540-74866540 on Build GRCh38
rs893285998 Chr.17: 74866564-74866564 on Build GRCh38
rs769528827 Chr.17: 74866414-74866414 on Build GRCh38
rs773160243 Chr.17: 74866420-74866420 on Build GRCh38
rs571288519 Chr.17: 74866486-74866486 on Build GRCh38
rs975722496 Chr.17: 74866389-74866389 on Build GRCh38
rs372003980 Chr.17: 74866571-74866571 on Build GRCh38
rs376876971 Chr.17: 74866575-74866575 on Build GRCh38
rs894527866 Chr.17: 74866484-74866484 on Build GRCh38
rs369049042 Chr.17: 74866480-74866480 on Build GRCh38
rs761931242 Chr.17: 74866533-74866533 on Build GRCh38
rs901108258 Chr.17: 74866531-74866531 on Build GRCh38
rs372205255 Chr.17: 74866429-74866429 on Build GRCh38
rs748969330 Chr.17: 74866509-74866509 on Build GRCh38
rs367813817 Chr.17: 74866525-74866525 on Build GRCh38

View Full Product Details