Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 721699-721936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM173A
Entrez Gene ID
65990
Gene Name
family with sequence similarity 173 member A
Gene Aliases
C16orf24
UniGene
Hs.166244
Gene Chromosome Location
Chr.16: 720581-722590 on Build GRCh38
Transcripts
NM_001271285.1 NM_023933.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7301829 c.276G>A p.A92A Chr.16: 721809-721809 on Build GRCh38
5570521 c.304G>A p.V102M Chr.16: 721837-721837 on Build GRCh38
4141910 c.394A>C p.K132Q Chr.16: 721927-721927 on Build GRCh38
7213407 c.275C>T p.A92V Chr.16: 721808-721808 on Build GRCh38
5484055 c.372C>T p.A124A Chr.16: 721905-721905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749347121 Chr.16: 721751-721751 on Build GRCh38
rs765111676 Chr.16: 721741-721741 on Build GRCh38
rs369258905 Chr.16: 721765-721765 on Build GRCh38
rs35763506 Chr.16: 721861-721862 on Build GRCh38
rs766556681 Chr.16: 721832-721832 on Build GRCh38
rs62032514 Chr.16: 721817-721817 on Build GRCh38
rs755078999 Chr.16: 721834-721834 on Build GRCh38
rs371628834 Chr.16: 721738-721738 on Build GRCh38
rs775083357 Chr.16: 721882-721882 on Build GRCh38
rs753294531 Chr.16: 721730-721730 on Build GRCh38
rs774405758 Chr.16: 721885-721885 on Build GRCh38
rs556451617 Chr.16: 721840-721840 on Build GRCh38
rs759019578 Chr.16: 721776-721776 on Build GRCh38
rs769250066 Chr.16: 721877-721877 on Build GRCh38
rs374348606 Chr.16: 721906-721906 on Build GRCh38
rs745533194 Chr.16: 721875-721875 on Build GRCh38
rs748683001 Chr.16: 721757-721757 on Build GRCh38
rs757065673 Chr.16: 721823-721823 on Build GRCh38
rs373177419 Chr.16: 721775-721775 on Build GRCh38
rs775360296 Chr.16: 721796-721796 on Build GRCh38
rs762974638 Chr.16: 721809-721809 on Build GRCh38
rs773579644 Chr.16: 721758-721758 on Build GRCh38
rs758171593 Chr.16: 721744-721744 on Build GRCh38
rs771265207 Chr.16: 721855-721855 on Build GRCh38
rs982910710 Chr.16: 721731-721731 on Build GRCh38
rs776188794 Chr.16: 721870-721870 on Build GRCh38
rs747307584 Chr.16: 721762-721762 on Build GRCh38
rs940417148 Chr.16: 721739-721739 on Build GRCh38
rs907631769 Chr.16: 721736-721736 on Build GRCh38
rs754064432 Chr.16: 721903-721903 on Build GRCh38
rs747397568 Chr.16: 721854-721854 on Build GRCh38
rs764051935 Chr.16: 721811-721811 on Build GRCh38
rs768757022 Chr.16: 721754-721754 on Build GRCh38
rs751428024 Chr.16: 721818-721818 on Build GRCh38
rs1025139657 Chr.16: 721786-721786 on Build GRCh38
rs779326230 Chr.16: 721755-721755 on Build GRCh38
rs999764399 Chr.16: 721824-721824 on Build GRCh38
rs754057848 Chr.16: 721833-721833 on Build GRCh38
rs777710654 Chr.16: 721746-721746 on Build GRCh38
rs370159029 Chr.16: 721884-721884 on Build GRCh38
rs756405709 Chr.16: 721749-721749 on Build GRCh38
rs778213366 Chr.16: 721851-721851 on Build GRCh38
rs748118024 Chr.16: 721841-721841 on Build GRCh38
rs761738482 Chr.16: 721890-721890 on Build GRCh38
rs767338108 Chr.16: 721901-721901 on Build GRCh38
rs1052230317 Chr.16: 721783-721783 on Build GRCh38
rs1032775585 Chr.16: 721837-721837 on Build GRCh38
rs3752555 Chr.16: 721750-721750 on Build GRCh38
rs764776808 Chr.16: 721789-721789 on Build GRCh38
rs752661408 Chr.16: 721743-721743 on Build GRCh38
rs750736654 Chr.16: 721748-721748 on Build GRCh38
rs902798089 Chr.16: 721822-721822 on Build GRCh38
rs1020097186 Chr.16: 721845-721845 on Build GRCh38

View Full Product Details