Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 161689583-161689832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRA6
Entrez Gene ID
2559
Gene Name
gamma-aminobutyric acid type A receptor alpha6 subunit
Gene Aliases
-
UniGene
Hs.90791
Gene Chromosome Location
Chr.5: 161685652-161702592 on Build GRCh38
Transcripts
NM_000811.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
139626 c.564G>A p.W188* Chr.5: 161689670-161689670 on Build GRCh38
7104204 c.530-8A>G p.? Chr.5: 161689628-161689628 on Build GRCh38
3715031 c.630G>T p.L210L Chr.5: 161689736-161689736 on Build GRCh38
5950794 c.564G>C p.W188C Chr.5: 161689670-161689670 on Build GRCh38
5023725 c.566A>T p.K189I Chr.5: 161689672-161689672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772826373 Chr.5: 161689714-161689714 on Build GRCh38
rs754107619 Chr.5: 161689753-161689753 on Build GRCh38
rs762734884 Chr.5: 161689610-161689610 on Build GRCh38
rs3811993 Chr.5: 161689666-161689666 on Build GRCh38
rs764380787 Chr.5: 161689639-161689639 on Build GRCh38
rs1042913786 Chr.5: 161689719-161689719 on Build GRCh38
rs200968260 Chr.5: 161689769-161689769 on Build GRCh38
rs752120603 Chr.5: 161689780-161689780 on Build GRCh38
rs147064708 Chr.5: 161689754-161689754 on Build GRCh38
rs752671384 Chr.5: 161689621-161689621 on Build GRCh38
rs368251992 Chr.5: 161689798-161689798 on Build GRCh38
rs367733338 Chr.5: 161689756-161689756 on Build GRCh38
rs762057483 Chr.5: 161689759-161689759 on Build GRCh38
rs762829158 Chr.5: 161689724-161689724 on Build GRCh38
rs200341002 Chr.5: 161689782-161689782 on Build GRCh38
rs34826485 Chr.5: 161689745-161689745 on Build GRCh38
rs760815978 Chr.5: 161689740-161689740 on Build GRCh38
rs765506471 Chr.5: 161689768-161689768 on Build GRCh38
rs541145520 Chr.5: 161689804-161689804 on Build GRCh38
rs760867520 Chr.5: 161689632-161689632 on Build GRCh38
rs774386607 Chr.5: 161689737-161689737 on Build GRCh38
rs574955747 Chr.5: 161689644-161689644 on Build GRCh38
rs768208279 Chr.5: 161689677-161689677 on Build GRCh38
rs748105776 Chr.5: 161689691-161689696 on Build GRCh38
rs900645002 Chr.5: 161689749-161689749 on Build GRCh38
rs183133713 Chr.5: 161689792-161689792 on Build GRCh38
rs768545890 Chr.5: 161689611-161689614 on Build GRCh38
rs780406601 Chr.5: 161689779-161689779 on Build GRCh38
rs747671960 Chr.5: 161689794-161689794 on Build GRCh38
rs138428207 Chr.5: 161689771-161689771 on Build GRCh38
rs757682550 Chr.5: 161689658-161689658 on Build GRCh38
rs868382842 Chr.5: 161689669-161689669 on Build GRCh38
rs928024636 Chr.5: 161689627-161689627 on Build GRCh38
rs145469537 Chr.5: 161689731-161689731 on Build GRCh38
rs747570774 Chr.5: 161689727-161689727 on Build GRCh38
rs200710979 Chr.5: 161689667-161689667 on Build GRCh38
rs780812038 Chr.5: 161689691-161689691 on Build GRCh38
rs143830239 Chr.5: 161689700-161689700 on Build GRCh38
rs769441572 Chr.5: 161689704-161689704 on Build GRCh38
rs779209499 Chr.5: 161689662-161689662 on Build GRCh38
rs754160838 Chr.5: 161689656-161689656 on Build GRCh38
rs925743412 Chr.5: 161689728-161689728 on Build GRCh38
rs766354679 Chr.5: 161689619-161689619 on Build GRCh38
rs774441339 Chr.5: 161689652-161689657 on Build GRCh38
rs900861590 Chr.5: 161689806-161689806 on Build GRCh38
rs770734349 Chr.5: 161689729-161689729 on Build GRCh38
rs746474290 Chr.5: 161689672-161689672 on Build GRCh38

View Full Product Details