Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 45026270-45026543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC3B
Entrez Gene ID
7123
Gene Name
C-type lectin domain family 3 member B
Gene Aliases
TN, TNA
UniGene
Hs.476092
Gene Chromosome Location
Chr.3: 45026207-45036073 on Build GRCh38
Transcripts
NM_003278.2 XM_017007116.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5603207 c.27C>T p.L9L Chr.3: 45026389-45026389 on Build GRCh38
4481344 c.24C>T p.L8L Chr.3: 45026386-45026386 on Build GRCh38
5712157 c.48G>C p.L16L Chr.3: 45026410-45026410 on Build GRCh38
4546418 c.3G>A p.M1I Chr.3: 45026365-45026365 on Build GRCh38
5819174 c.5A>G p.E2G Chr.3: 45026367-45026367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752188377 Chr.3: 45026474-45026476 on Build GRCh38
rs750319240 Chr.3: 45026368-45026368 on Build GRCh38
rs749574822 Chr.3: 45026405-45026405 on Build GRCh38
rs374914988 Chr.3: 45026407-45026407 on Build GRCh38
rs948874142 Chr.3: 45026297-45026297 on Build GRCh38
rs779081201 Chr.3: 45026517-45026517 on Build GRCh38
rs746488621 Chr.3: 45026326-45026326 on Build GRCh38
rs767886360 Chr.3: 45026353-45026353 on Build GRCh38
rs182424620 Chr.3: 45026348-45026348 on Build GRCh38
rs754708053 Chr.3: 45026508-45026508 on Build GRCh38
rs562246387 Chr.3: 45026352-45026352 on Build GRCh38
rs764766321 Chr.3: 45026363-45026363 on Build GRCh38
rs748351681 Chr.3: 45026346-45026346 on Build GRCh38
rs761483605 Chr.3: 45026484-45026484 on Build GRCh38
rs771193124 Chr.3: 45026350-45026350 on Build GRCh38
rs772459032 Chr.3: 45026413-45026413 on Build GRCh38
rs776234984 Chr.3: 45026476-45026476 on Build GRCh38
rs780593892 Chr.3: 45026492-45026492 on Build GRCh38
rs572305560 Chr.3: 45026386-45026386 on Build GRCh38
rs898530456 Chr.3: 45026318-45026318 on Build GRCh38
rs940147991 Chr.3: 45026310-45026310 on Build GRCh38
rs760260623 Chr.3: 45026516-45026517 on Build GRCh38
rs763645318 Chr.3: 45026356-45026356 on Build GRCh38
rs753288792 Chr.3: 45026321-45026321 on Build GRCh38
rs981641822 Chr.3: 45026308-45026308 on Build GRCh38
rs779169124 Chr.3: 45026408-45026408 on Build GRCh38
rs767131313 Chr.3: 45026383-45026385 on Build GRCh38
rs779743973 Chr.3: 45026338-45026338 on Build GRCh38
rs779841981 Chr.3: 45026377-45026377 on Build GRCh38
rs201995829 Chr.3: 45026515-45026515 on Build GRCh38
rs928673241 Chr.3: 45026309-45026309 on Build GRCh38
rs2301281 Chr.3: 45026332-45026332 on Build GRCh38
rs745936314 Chr.3: 45026521-45026521 on Build GRCh38
rs746331675 Chr.3: 45026410-45026410 on Build GRCh38
rs1048985371 Chr.3: 45026314-45026314 on Build GRCh38
rs754077114 Chr.3: 45026513-45026513 on Build GRCh38
rs751317349 Chr.3: 45026378-45026378 on Build GRCh38
rs762884123 Chr.3: 45026497-45026497 on Build GRCh38
rs534091806 Chr.3: 45026425-45026425 on Build GRCh38
rs781085423 Chr.3: 45026509-45026509 on Build GRCh38
rs758352607 Chr.3: 45026374-45026374 on Build GRCh38
rs761291183 Chr.3: 45026361-45026361 on Build GRCh38
rs745855000 Chr.3: 45026330-45026330 on Build GRCh38
rs113082596 Chr.3: 45026496-45026496 on Build GRCh38
rs749374562 Chr.3: 45026349-45026349 on Build GRCh38
rs769108845 Chr.3: 45026436-45026436 on Build GRCh38
rs777844887 Chr.3: 45026394-45026394 on Build GRCh38
rs34138982 Chr.3: 45026359-45026359 on Build GRCh38
rs953151106 Chr.3: 45026505-45026505 on Build GRCh38
rs373086753 Chr.3: 45026435-45026435 on Build GRCh38
rs200880858 Chr.3: 45026315-45026315 on Build GRCh38
rs754870570 Chr.3: 45026385-45026385 on Build GRCh38
rs369932221 Chr.3: 45026360-45026360 on Build GRCh38
rs751572880 Chr.3: 45026506-45026506 on Build GRCh38
rs766224879 Chr.3: 45026499-45026499 on Build GRCh38
rs778804129 Chr.3: 45026328-45026328 on Build GRCh38
rs760306489 Chr.3: 45026354-45026354 on Build GRCh38

View Full Product Details