Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 14747203-14747462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FREM1
Entrez Gene ID
158326
Gene Name
FRAS1 related extracellular matrix 1
Gene Aliases
BNAR, C9orf143, C9orf145, C9orf154, MOTA, TILRR, TRIGNO2
UniGene
Hs.50850
Gene Chromosome Location
Chr.9: 14734666-14911653 on Build GRCh38
Transcripts
NM_001177704.1 NM_144966.5 XM_005251382.3 XM_005251384.4 XM_006716729.3 XM_011517758.2 XM_017014316.1 XM_017014317.1 XM_017014318.1 XM_017014319.1 XM_017014320.1 XM_017014321.1 XM_017014322.1 XM_017014323.1 XM_017014324.1 XM_017014325.1 XM_017014326.1 XM_017014327.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7237696 c.5873A>G p.E1958G Chr.9: 14747400-14747400 on Build GRCh38
6033843 c.1493T>G p.F498C Chr.9: 14747388-14747388 on Build GRCh38
7368770 c.1552A>G p.K518E Chr.9: 14747329-14747329 on Build GRCh38
6901076 c.6004A>G p.R2002G Chr.9: 14747269-14747269 on Build GRCh38
6183030 c.5989G>A p.D1997N Chr.9: 14747284-14747284 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs4741426 Chr.9: 14747414-14747414 on Build GRCh38
rs745524714 Chr.9: 14747285-14747285 on Build GRCh38
rs777744025 Chr.9: 14747260-14747260 on Build GRCh38
rs763985268 Chr.9: 14747369-14747369 on Build GRCh38
rs754058350 Chr.9: 14747233-14747233 on Build GRCh38
rs759680427 Chr.9: 14747355-14747355 on Build GRCh38
rs772616034 Chr.9: 14747396-14747396 on Build GRCh38
rs769002310 Chr.9: 14747307-14747307 on Build GRCh38
rs773896918 Chr.9: 14747329-14747329 on Build GRCh38
rs780594544 Chr.9: 14747383-14747383 on Build GRCh38
rs375849105 Chr.9: 14747399-14747399 on Build GRCh38
rs201002953 Chr.9: 14747419-14747419 on Build GRCh38
rs769901416 Chr.9: 14747426-14747426 on Build GRCh38
rs756797204 Chr.9: 14747271-14747271 on Build GRCh38
rs759720397 Chr.9: 14747421-14747421 on Build GRCh38
rs1049876248 Chr.9: 14747397-14747397 on Build GRCh38
rs758886562 Chr.9: 14747357-14747357 on Build GRCh38
rs760818080 Chr.9: 14747330-14747330 on Build GRCh38
rs368187885 Chr.9: 14747328-14747328 on Build GRCh38
rs765024237 Chr.9: 14747356-14747356 on Build GRCh38
rs779162732 Chr.9: 14747309-14747309 on Build GRCh38
rs756703645 Chr.9: 14747381-14747381 on Build GRCh38
rs564553123 Chr.9: 14747234-14747234 on Build GRCh38
rs886500882 Chr.9: 14747256-14747256 on Build GRCh38
rs752419764 Chr.9: 14747362-14747362 on Build GRCh38
rs766678522 Chr.9: 14747230-14747230 on Build GRCh38
rs779825488 Chr.9: 14747388-14747388 on Build GRCh38
rs763196871 Chr.9: 14747234-14747237 on Build GRCh38
rs977729728 Chr.9: 14747367-14747367 on Build GRCh38
rs1026367640 Chr.9: 14747238-14747238 on Build GRCh38
rs755712880 Chr.9: 14747387-14747387 on Build GRCh38
rs1006346420 Chr.9: 14747258-14747258 on Build GRCh38
rs375669260 Chr.9: 14747317-14747317 on Build GRCh38
rs549558249 Chr.9: 14747409-14747409 on Build GRCh38
rs780797923 Chr.9: 14747282-14747282 on Build GRCh38
rs549309618 Chr.9: 14747408-14747408 on Build GRCh38
rs968813593 Chr.9: 14747320-14747320 on Build GRCh38
rs200035180 Chr.9: 14747235-14747235 on Build GRCh38
rs751544988 Chr.9: 14747380-14747380 on Build GRCh38
rs978438400 Chr.9: 14747324-14747324 on Build GRCh38
rs776873418 Chr.9: 14747354-14747354 on Build GRCh38
rs752612632 Chr.9: 14747245-14747245 on Build GRCh38
rs758344694 Chr.9: 14747253-14747253 on Build GRCh38
rs199497460 Chr.9: 14747364-14747364 on Build GRCh38
rs183882265 Chr.9: 14747331-14747331 on Build GRCh38
rs370544270 Chr.9: 14747313-14747313 on Build GRCh38
rs748496318 Chr.9: 14747391-14747391 on Build GRCh38
rs760571146 Chr.9: 14747323-14747323 on Build GRCh38
rs746516776 Chr.9: 14747268-14747268 on Build GRCh38

View Full Product Details