Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 94553931-94554168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HELLS
Entrez Gene ID
3070
Gene Name
helicase, lymphoid-specific
Gene Aliases
ICF4, LSH, Nbla10143, PASG, SMARCA6
UniGene
Hs.463677
Gene Chromosome Location
Chr.10: 94545767-94613905 on Build GRCh38
Transcripts
NM_001289067.1 NM_001289068.1 NM_001289069.1 NM_001289070.1 NM_001289071.1 NM_001289072.1 NM_001289073.1 NM_001289074.1 NM_001289075.1 NM_018063.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4972347 c.190C>T p.R64W Chr.10: 94554162-94554162 on Build GRCh38
4016853 c.193T>C p.Y65H Chr.10: 94554165-94554165 on Build GRCh38
240096 c.190C>T p.R64W Chr.10: 94554162-94554162 on Build GRCh38
6678985 c.196C>T p.R66C Chr.10: 94554168-94554168 on Build GRCh38
5178578 c.158G>A p.R53H Chr.10: 94554130-94554130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747283119 Chr.10: 94554132-94554132 on Build GRCh38
rs1032589716 Chr.10: 94554092-94554092 on Build GRCh38
rs750777864 Chr.10: 94554124-94554124 on Build GRCh38
rs766960365 Chr.10: 94554100-94554100 on Build GRCh38
rs1013308202 Chr.10: 94554037-94554045 on Build GRCh38
rs61547368 Chr.10: 94553983-94553984 on Build GRCh38
rs966420466 Chr.10: 94554059-94554059 on Build GRCh38
rs576686304 Chr.10: 94554099-94554099 on Build GRCh38
rs781134124 Chr.10: 94554142-94554142 on Build GRCh38
rs751295310 Chr.10: 94554097-94554097 on Build GRCh38
rs141169560 Chr.10: 94554129-94554129 on Build GRCh38
rs894773596 Chr.10: 94554019-94554019 on Build GRCh38
rs191523820 Chr.10: 94554008-94554008 on Build GRCh38
rs375345457 Chr.10: 94553973-94553974 on Build GRCh38
rs531049416 Chr.10: 94554086-94554088 on Build GRCh38
rs899454258 Chr.10: 94553998-94553998 on Build GRCh38
rs116632313 Chr.10: 94553987-94553987 on Build GRCh38
rs543332826 Chr.10: 94554067-94554067 on Build GRCh38
rs1002186083 Chr.10: 94554002-94554002 on Build GRCh38
rs940866444 Chr.10: 94553974-94553974 on Build GRCh38
rs556828408 Chr.10: 94554078-94554078 on Build GRCh38
rs757709828 Chr.10: 94554108-94554108 on Build GRCh38
rs766124607 Chr.10: 94554091-94554091 on Build GRCh38
rs1025551640 Chr.10: 94554093-94554093 on Build GRCh38
rs748057339 Chr.10: 94554143-94554143 on Build GRCh38
rs574256065 Chr.10: 94554018-94554018 on Build GRCh38
rs754331856 Chr.10: 94554106-94554106 on Build GRCh38
rs762897748 Chr.10: 94554081-94554081 on Build GRCh38
rs780221772 Chr.10: 94554130-94554130 on Build GRCh38
rs768738765 Chr.10: 94554139-94554139 on Build GRCh38
rs779280223 Chr.10: 94554122-94554122 on Build GRCh38
rs373332345 Chr.10: 94554047-94554047 on Build GRCh38

View Full Product Details