Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18744495-18744722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW10
Entrez Gene ID
10517
Gene Name
F-box and WD repeat domain containing 10
Gene Aliases
Fbw10, HREP, SM25H2, SM2SH2
UniGene
Hs.592128
Gene Chromosome Location
Chr.17: 18744013-18779349 on Build GRCh38
Transcripts
NM_001267585.1 NM_001267586.1 NR_051988.1 XM_011523625.1 XM_011523626.2 XM_017024020.1 XM_017024021.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1177435 c.273G>A p.R91R Chr.17: 18744517-18744517 on Build GRCh38
7140016 c.272G>T p.R91L Chr.17: 18744516-18744516 on Build GRCh38
7140017 c.272G>T p.R91L Chr.17: 18744516-18744516 on Build GRCh38
5959621 c.277A>G p.N93D Chr.17: 18744521-18744521 on Build GRCh38
4579501 c.408T>C p.Y136Y Chr.17: 18744652-18744652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760016525 Chr.17: 18744620-18744620 on Build GRCh38
rs768357844 Chr.17: 18744590-18744590 on Build GRCh38
rs753966727 Chr.17: 18744628-18744628 on Build GRCh38
rs145973790 Chr.17: 18744652-18744652 on Build GRCh38
rs747613611 Chr.17: 18744609-18744609 on Build GRCh38
rs774691557 Chr.17: 18744614-18744614 on Build GRCh38
rs758298921 Chr.17: 18744646-18744646 on Build GRCh38
rs760915440 Chr.17: 18744625-18744625 on Build GRCh38
rs377476079 Chr.17: 18744548-18744548 on Build GRCh38
rs778797983 Chr.17: 18744558-18744558 on Build GRCh38
rs531699302 Chr.17: 18744613-18744613 on Build GRCh38
rs764249402 Chr.17: 18744626-18744626 on Build GRCh38
rs765225589 Chr.17: 18744635-18744635 on Build GRCh38
rs764159611 Chr.17: 18744547-18744547 on Build GRCh38
rs758122859 Chr.17: 18744561-18744561 on Build GRCh38
rs142528410 Chr.17: 18744587-18744587 on Build GRCh38
rs779717723 Chr.17: 18744651-18744651 on Build GRCh38
rs757205842 Chr.17: 18744556-18744556 on Build GRCh38
rs756298001 Chr.17: 18744530-18744530 on Build GRCh38
rs573539157 Chr.17: 18744535-18744536 on Build GRCh38
rs775852531 Chr.17: 18744622-18744622 on Build GRCh38
rs754700744 Chr.17: 18744674-18744674 on Build GRCh38
rs750314402 Chr.17: 18744645-18744645 on Build GRCh38
rs772332798 Chr.17: 18744621-18744621 on Build GRCh38
rs965011265 Chr.17: 18744578-18744578 on Build GRCh38
rs779813238 Chr.17: 18744566-18744566 on Build GRCh38
rs778033611 Chr.17: 18744654-18744655 on Build GRCh38
rs780972225 Chr.17: 18744681-18744681 on Build GRCh38
rs757856483 Chr.17: 18744675-18744676 on Build GRCh38
rs780882213 Chr.17: 18744595-18744595 on Build GRCh38
rs745713216 Chr.17: 18744560-18744560 on Build GRCh38
rs761849181 Chr.17: 18744630-18744630 on Build GRCh38

View Full Product Details