Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32737205-32737432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAXBP1
Entrez Gene ID
94104
Gene Name
PAX3 and PAX7 binding protein 1
Gene Aliases
BM020, C21orf66, FSAP105, GCFC, GCFC1
UniGene
Hs.644004
Gene Chromosome Location
Chr.21: 32733899-32771858 on Build GRCh38
Transcripts
NM_016631.3 NR_027873.1 XM_006724066.1 NR_038879.1

Gene Symbol
PAXBP1-AS1
Entrez Gene ID
100506215
Gene Name
PAXBP1 antisense RNA 1
Gene Aliases
GCFC1-AS1
UniGene
Hs.657123
Gene Chromosome Location
Chr.21: 32728114-32743122 on Build GRCh38
Transcripts
NM_016631.3 NR_027873.1 XM_006724066.1 NR_038879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7189888 c.2610_2611insA p.S871fs*2 Chr.21: 32737279-32737280 on Build GRCh38
6738315 c.2591A>C p.Y864S Chr.21: 32737299-32737299 on Build GRCh38
6306220 c.2633C>A p.A878E Chr.21: 32737257-32737257 on Build GRCh38
1161403 c.2569G>A p.V857I Chr.21: 32737321-32737321 on Build GRCh38
1030240 c.2505A>C p.Q835H Chr.21: 32737385-32737385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775040197 Chr.21: 32737376-32737376 on Build GRCh38
rs745722467 Chr.21: 32737349-32737349 on Build GRCh38
rs551236608 Chr.21: 32737295-32737295 on Build GRCh38
rs563031399 Chr.21: 32737304-32737304 on Build GRCh38
rs763482590 Chr.21: 32737293-32737293 on Build GRCh38
rs749435435 Chr.21: 32737234-32737234 on Build GRCh38
rs370603447 Chr.21: 32737283-32737283 on Build GRCh38
rs770259842 Chr.21: 32737321-32737321 on Build GRCh38
rs760246998 Chr.21: 32737286-32737286 on Build GRCh38
rs775447295 Chr.21: 32737394-32737394 on Build GRCh38
rs1049444035 Chr.21: 32737307-32737307 on Build GRCh38
rs370348660 Chr.21: 32737401-32737401 on Build GRCh38
rs562272048 Chr.21: 32737253-32737253 on Build GRCh38
rs138511262 Chr.21: 32737329-32737329 on Build GRCh38
rs144102538 Chr.21: 32737403-32737403 on Build GRCh38
rs770804239 Chr.21: 32737242-32737242 on Build GRCh38
rs755335296 Chr.21: 32737317-32737317 on Build GRCh38
rs147145324 Chr.21: 32737319-32737319 on Build GRCh38
rs779053019 Chr.21: 32737347-32737347 on Build GRCh38
rs199811323 Chr.21: 32737334-32737334 on Build GRCh38
rs751976064 Chr.21: 32737310-32737310 on Build GRCh38
rs376912985 Chr.21: 32737365-32737365 on Build GRCh38
rs199655078 Chr.21: 32737257-32737257 on Build GRCh38
rs199507974 Chr.21: 32737330-32737330 on Build GRCh38
rs749337356 Chr.21: 32737332-32737332 on Build GRCh38
rs774050399 Chr.21: 32737247-32737247 on Build GRCh38
rs867702535 Chr.21: 32737285-32737285 on Build GRCh38
rs143382738 Chr.21: 32737271-32737271 on Build GRCh38

View Full Product Details