Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 42087855-42088082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GXYLT1
Entrez Gene ID
283464
Gene Name
glucoside xylosyltransferase 1
Gene Aliases
GLT8D3
UniGene
Hs.259347
Gene Chromosome Location
Chr.12: 42081845-42144871 on Build GRCh38
Transcripts
NM_001099650.1 NM_173601.1 XM_017019211.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1731351 c.1241G>A p.C414Y Chr.12: 42087868-42087868 on Build GRCh38
5739614 c.1225_1226insA p.T409fs*>33 Chr.12: 42087883-42087884 on Build GRCh38
1731354 c.1147T>C p.C383R Chr.12: 42087869-42087869 on Build GRCh38
7214823 c.1086C>T p.D362D Chr.12: 42087930-42087930 on Build GRCh38
1731353 c.1240T>C p.C414R Chr.12: 42087869-42087869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190668767 Chr.12: 42087966-42087966 on Build GRCh38
rs140041049 Chr.12: 42088004-42088005 on Build GRCh38
rs751301325 Chr.12: 42087905-42087905 on Build GRCh38
rs200436402 Chr.12: 42087886-42087886 on Build GRCh38
rs824722 Chr.12: 42088001-42088001 on Build GRCh38
rs754185733 Chr.12: 42087926-42087926 on Build GRCh38
rs761317050 Chr.12: 42087991-42087991 on Build GRCh38
rs755383414 Chr.12: 42087928-42087928 on Build GRCh38
rs772359758 Chr.12: 42087943-42087943 on Build GRCh38
rs371166830 Chr.12: 42087957-42087957 on Build GRCh38
rs760286336 Chr.12: 42087996-42087996 on Build GRCh38
rs887225421 Chr.12: 42087952-42087952 on Build GRCh38
rs758734999 Chr.12: 42087972-42087972 on Build GRCh38
rs939393048 Chr.12: 42087923-42087923 on Build GRCh38
rs768535402 Chr.12: 42087983-42087983 on Build GRCh38
rs750671452 Chr.12: 42087965-42087966 on Build GRCh38
rs987745897 Chr.12: 42088047-42088047 on Build GRCh38
rs7972986 Chr.12: 42087930-42087930 on Build GRCh38
rs752874350 Chr.12: 42088012-42088012 on Build GRCh38
rs573606046 Chr.12: 42088000-42088000 on Build GRCh38
rs761561507 Chr.12: 42087908-42087908 on Build GRCh38
rs749960332 Chr.12: 42087917-42087917 on Build GRCh38
rs774313510 Chr.12: 42087987-42087987 on Build GRCh38
rs780599778 Chr.12: 42087924-42087924 on Build GRCh38
rs759764940 Chr.12: 42087972-42087972 on Build GRCh38
rs747183028 Chr.12: 42087965-42087965 on Build GRCh38
rs748327835 Chr.12: 42087941-42087941 on Build GRCh38
rs749894191 Chr.12: 42087995-42087995 on Build GRCh38
rs558061830 Chr.12: 42087922-42087922 on Build GRCh38
rs138027653 Chr.12: 42088041-42088041 on Build GRCh38
rs767200986 Chr.12: 42087993-42087993 on Build GRCh38
rs555254165 Chr.12: 42087931-42087931 on Build GRCh38
rs561828822 Chr.12: 42087997-42087997 on Build GRCh38
rs1018635641 Chr.12: 42088018-42088018 on Build GRCh38
rs766891142 Chr.12: 42087914-42087914 on Build GRCh38

View Full Product Details