Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46934888-46935127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOSR2
Entrez Gene ID
9570
Gene Name
golgi SNAP receptor complex member 2
Gene Aliases
Bos1, EPM6, GS27
UniGene
Hs.463278
Gene Chromosome Location
Chr.17: 46923086-46975519 on Build GRCh38
Transcripts
NM_001012511.2 NM_001321133.1 NM_001321134.1 NM_004287.4 NM_054022.3 XM_005257844.3 XM_006722190.3 XM_011525501.2 XM_011525502.2 XM_017025378.1 XM_017025379.1 XM_017025380.1 XM_017025381.1 XM_017025382.1 XM_017025383.1 XM_017025384.1 XM_017025385.1 XM_017025386.1 XM_017025387.1 XM_017025388.1 XM_017025389.1 XM_017025390.1 XM_017025391.1 XM_017025392.1 XM_017025393.1 XM_017025394.1 XM_017025395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6022754 c.365A>G p.E122G Chr.17: 46935057-46935057 on Build GRCh38
7237078 c.337-4A>T p.? Chr.17: 46935025-46935025 on Build GRCh38
6022755 c.365A>G p.E122G Chr.17: 46935057-46935057 on Build GRCh38
3518437 c.384C>T p.S128S Chr.17: 46935076-46935076 on Build GRCh38
5852760 c.412G>A p.D138N Chr.17: 46935104-46935104 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761272920 Chr.17: 46935073-46935073 on Build GRCh38
rs768238866 Chr.17: 46935067-46935067 on Build GRCh38
rs926792258 Chr.17: 46934947-46934947 on Build GRCh38
rs757420754 Chr.17: 46935035-46935035 on Build GRCh38
rs563155623 Chr.17: 46935083-46935083 on Build GRCh38
rs560950648 Chr.17: 46934935-46934935 on Build GRCh38
rs765513191 Chr.17: 46935096-46935096 on Build GRCh38
rs780939347 Chr.17: 46934990-46934990 on Build GRCh38
rs759767385 Chr.17: 46934994-46934994 on Build GRCh38
rs770194081 Chr.17: 46934931-46934931 on Build GRCh38
rs939504353 Chr.17: 46934976-46934976 on Build GRCh38
rs751635119 Chr.17: 46935032-46935032 on Build GRCh38
rs909664511 Chr.17: 46934921-46934921 on Build GRCh38
rs777988995 Chr.17: 46935020-46935020 on Build GRCh38
rs137975476 Chr.17: 46935080-46935080 on Build GRCh38
rs781240194 Chr.17: 46935040-46935040 on Build GRCh38
rs542483245 Chr.17: 46934930-46934930 on Build GRCh38
rs747784735 Chr.17: 46935017-46935017 on Build GRCh38
rs150381512 Chr.17: 46935061-46935061 on Build GRCh38
rs746266977 Chr.17: 46934914-46934914 on Build GRCh38
rs745840057 Chr.17: 46935044-46935044 on Build GRCh38
rs886053078 Chr.17: 46935055-46935055 on Build GRCh38
rs866215225 Chr.17: 46934926-46934926 on Build GRCh38
rs763426536 Chr.17: 46935100-46935100 on Build GRCh38
rs752828696 Chr.17: 46935007-46935007 on Build GRCh38
rs374613085 Chr.17: 46935056-46935056 on Build GRCh38
rs182039780 Chr.17: 46935085-46935085 on Build GRCh38
rs766803823 Chr.17: 46934978-46934978 on Build GRCh38
rs773995951 Chr.17: 46935072-46935072 on Build GRCh38
rs754400864 Chr.17: 46934980-46934980 on Build GRCh38
rs758711013 Chr.17: 46935019-46935019 on Build GRCh38
rs753166413 Chr.17: 46935097-46935097 on Build GRCh38
rs765679030 Chr.17: 46934996-46934996 on Build GRCh38
rs769454681 Chr.17: 46935050-46935050 on Build GRCh38
rs140105943 Chr.17: 46935075-46935075 on Build GRCh38
rs529781220 Chr.17: 46934965-46934965 on Build GRCh38

View Full Product Details