Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 105471755-105472010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSD1L
Entrez Gene ID
83856
Gene Name
fibronectin type III and SPRY domain containing 1 like
Gene Aliases
CCDC10, CSDUFD1, FSD1CL, FSD1NL, MIR1
UniGene
Hs.136901
Gene Chromosome Location
Chr.9: 105442183-105552433 on Build GRCh38
Transcripts
NM_001145313.2 NM_001287191.1 NM_001287192.1 NM_031919.4 XM_005252254.2 XM_005252257.3 XM_005252260.3 XM_011519077.2 XM_011519078.2 XM_011519079.2 XM_017015182.1 XM_017015183.1 XM_017015184.1 XM_017015185.1 XM_017015186.1 XM_017015187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5063476 c.367C>A p.L123M Chr.9: 105471931-105471931 on Build GRCh38
6315166 c.316G>T p.D106Y Chr.9: 105471976-105471976 on Build GRCh38
1624387 c.411A>G p.L137L Chr.9: 105471975-105471975 on Build GRCh38
6315165 c.412G>T p.D138Y Chr.9: 105471976-105471976 on Build GRCh38
1624388 c.315A>G p.L105L Chr.9: 105471975-105471975 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976615097 Chr.9: 105471870-105471870 on Build GRCh38
rs750203459 Chr.9: 105471898-105471898 on Build GRCh38
rs571540973 Chr.9: 105471919-105471919 on Build GRCh38
rs534688297 Chr.9: 105471838-105471838 on Build GRCh38
rs567948516 Chr.9: 105471867-105471867 on Build GRCh38
rs534875055 Chr.9: 105471879-105471879 on Build GRCh38
rs781418038 Chr.9: 105471905-105471905 on Build GRCh38
rs11421948 Chr.9: 105471897-105471898 on Build GRCh38
rs758289792 Chr.9: 105471900-105471900 on Build GRCh38
rs1006977118 Chr.9: 105471853-105471853 on Build GRCh38
rs61996264 Chr.9: 105471972-105471972 on Build GRCh38
rs1056200139 Chr.9: 105471979-105471979 on Build GRCh38
rs978758329 Chr.9: 105471847-105471847 on Build GRCh38
rs909535937 Chr.9: 105471951-105471951 on Build GRCh38
rs181585378 Chr.9: 105471957-105471957 on Build GRCh38
rs765094153 Chr.9: 105471897-105471897 on Build GRCh38
rs951311773 Chr.9: 105471881-105471881 on Build GRCh38
rs984029835 Chr.9: 105471942-105471942 on Build GRCh38
rs1021648207 Chr.9: 105471795-105471795 on Build GRCh38
rs766881535 Chr.9: 105471930-105471930 on Build GRCh38
rs28503319 Chr.9: 105471926-105471926 on Build GRCh38
rs752865462 Chr.9: 105471909-105471909 on Build GRCh38
rs775587678 Chr.9: 105471882-105471883 on Build GRCh38
rs770801403 Chr.9: 105471883-105471884 on Build GRCh38
rs555345664 Chr.9: 105471965-105471965 on Build GRCh38
rs771620631 Chr.9: 105471883-105471883 on Build GRCh38
rs779425076 Chr.9: 105471968-105471968 on Build GRCh38
rs753803025 Chr.9: 105471894-105471894 on Build GRCh38
rs557473739 Chr.9: 105471931-105471931 on Build GRCh38
rs1022212337 Chr.9: 105471802-105471802 on Build GRCh38
rs117347201 Chr.9: 105471961-105471961 on Build GRCh38
rs773058053 Chr.9: 105471805-105471805 on Build GRCh38
rs967922014 Chr.9: 105471822-105471822 on Build GRCh38
rs781277628 Chr.9: 105471848-105471848 on Build GRCh38
rs1018394376 Chr.9: 105471854-105471854 on Build GRCh38
rs553545599 Chr.9: 105471882-105471882 on Build GRCh38
rs757314499 Chr.9: 105471896-105471896 on Build GRCh38

View Full Product Details