Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 23000886-23001121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX53
Entrez Gene ID
168400
Gene Name
DEAD-box helicase 53
Gene Aliases
CAGE, CT26
UniGene
Hs.434416
Gene Chromosome Location
Chr.X: 22999961-23002089 on Build GRCh38
Transcripts
NM_182699.3 NR_073010.2

Gene Symbol
PTCHD1-AS
Entrez Gene ID
100873065
Gene Name
PTCHD1 antisense RNA (head to head)
Gene Aliases
DDX53-AS1, PTCHD1AS1, PTCHD1AS2
UniGene
Hs.628459
Gene Chromosome Location
Chr.X: 22193005-23293146 on Build GRCh38
Transcripts
NM_182699.3 NR_073010.2

SNP Detail

SNP ID Chromosome Location
rs753152349 Chr.X: 23001074-23001074 on Build GRCh38
rs781493806 Chr.X: 23001037-23001037 on Build GRCh38
rs758082063 Chr.X: 23001030-23001030 on Build GRCh38
rs917193512 Chr.X: 23000922-23000922 on Build GRCh38
rs937312541 Chr.X: 23000975-23000975 on Build GRCh38
rs371534137 Chr.X: 23000999-23000999 on Build GRCh38
rs932166391 Chr.X: 23001086-23001086 on Build GRCh38
rs368652147 Chr.X: 23001067-23001067 on Build GRCh38
rs756147094 Chr.X: 23000991-23000991 on Build GRCh38
rs774946901 Chr.X: 23000965-23000965 on Build GRCh38
rs771593872 Chr.X: 23001065-23001065 on Build GRCh38
rs750265276 Chr.X: 23001011-23001011 on Build GRCh38
rs778044990 Chr.X: 23001055-23001055 on Build GRCh38
rs767889537 Chr.X: 23000998-23000998 on Build GRCh38
rs760237113 Chr.X: 23000976-23000976 on Build GRCh38
rs763660535 Chr.X: 23000977-23000977 on Build GRCh38
rs748256605 Chr.X: 23001041-23001041 on Build GRCh38
rs772258905 Chr.X: 23001078-23001078 on Build GRCh38
rs767219682 Chr.X: 23000951-23000951 on Build GRCh38
rs866252322 Chr.X: 23000997-23000997 on Build GRCh38
rs142798320 Chr.X: 23001042-23001042 on Build GRCh38
rs372151229 Chr.X: 23001087-23001087 on Build GRCh38
rs967302685 Chr.X: 23000990-23000990 on Build GRCh38
rs759777747 Chr.X: 23000945-23000945 on Build GRCh38
rs368003698 Chr.X: 23000918-23000918 on Build GRCh38
rs373622053 Chr.X: 23001061-23001061 on Build GRCh38
rs753333307 Chr.X: 23000978-23000978 on Build GRCh38

View Full Product Details