Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41278992-41279265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPUL1
Entrez Gene ID
11100
Gene Name
heterogeneous nuclear ribonucleoprotein U like 1
Gene Aliases
E1B-AP5, E1BAP5, HNRPUL1
UniGene
Hs.155218
Gene Chromosome Location
Chr.19: 41262476-41307783 on Build GRCh38
Transcripts
NM_001301016.2 NM_001321208.1 NM_001321211.1 NM_007040.5 NM_144732.4 XM_005258459.3 XM_005258461.4 XM_005258463.3 XM_005258464.3 XM_005258465.4 XM_011526392.2 XM_011526393.2 XM_011526394.2 XM_011526395.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6681439 c.804C>T p.S268S Chr.19: 41279094-41279094 on Build GRCh38
4612098 c.834delC p.H280fs*16 Chr.19: 41279124-41279124 on Build GRCh38
6681444 c.832G>C p.D278H Chr.19: 41279122-41279122 on Build GRCh38
3534378 c.836C>T p.P279L Chr.19: 41279126-41279126 on Build GRCh38
712794 c.847C>T p.R283C Chr.19: 41279137-41279137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761718822 Chr.19: 41279156-41279156 on Build GRCh38
rs776048925 Chr.19: 41279150-41279150 on Build GRCh38
rs953380702 Chr.19: 41279100-41279100 on Build GRCh38
rs771875842 Chr.19: 41279030-41279030 on Build GRCh38
rs750270062 Chr.19: 41279081-41279081 on Build GRCh38
rs751525863 Chr.19: 41279109-41279109 on Build GRCh38
rs267605500 Chr.19: 41279136-41279136 on Build GRCh38
rs138404450 Chr.19: 41279115-41279115 on Build GRCh38
rs372122098 Chr.19: 41279130-41279130 on Build GRCh38
rs773985185 Chr.19: 41279149-41279153 on Build GRCh38
rs1016665676 Chr.19: 41279220-41279220 on Build GRCh38
rs758565657 Chr.19: 41279094-41279094 on Build GRCh38
rs755098689 Chr.19: 41279113-41279113 on Build GRCh38
rs765178808 Chr.19: 41279160-41279160 on Build GRCh38
rs749704371 Chr.19: 41279140-41279140 on Build GRCh38
rs772949221 Chr.19: 41279176-41279176 on Build GRCh38
rs764258898 Chr.19: 41279206-41279206 on Build GRCh38
rs771787658 Chr.19: 41279142-41279142 on Build GRCh38
rs748680952 Chr.19: 41279119-41279119 on Build GRCh38
rs781430477 Chr.19: 41279027-41279027 on Build GRCh38
rs751793371 Chr.19: 41279201-41279201 on Build GRCh38
rs760241098 Chr.19: 41279031-41279031 on Build GRCh38
rs1007571682 Chr.19: 41279194-41279194 on Build GRCh38
rs564582875 Chr.19: 41279019-41279019 on Build GRCh38
rs1005008485 Chr.19: 41279022-41279022 on Build GRCh38
rs914361323 Chr.19: 41279127-41279127 on Build GRCh38
rs889836288 Chr.19: 41279188-41279188 on Build GRCh38
rs963686331 Chr.19: 41279244-41279244 on Build GRCh38
rs373195309 Chr.19: 41279198-41279198 on Build GRCh38
rs776618619 Chr.19: 41279054-41279054 on Build GRCh38
rs761723830 Chr.19: 41279070-41279070 on Build GRCh38
rs180964950 Chr.19: 41279145-41279145 on Build GRCh38
rs79851033 Chr.19: 41279149-41279149 on Build GRCh38
rs766617661 Chr.19: 41279104-41279104 on Build GRCh38
rs756724566 Chr.19: 41279128-41279128 on Build GRCh38
rs762738046 Chr.19: 41279195-41279195 on Build GRCh38
rs1052304977 Chr.19: 41279036-41279036 on Build GRCh38
rs763884615 Chr.19: 41279050-41279050 on Build GRCh38
rs764986623 Chr.19: 41279079-41279079 on Build GRCh38

View Full Product Details