Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 62358084-62358353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF2A
Entrez Gene ID
3796
Gene Name
kinesin family member 2A
Gene Aliases
CDCBM3, HK2, KIF2
UniGene
Hs.558351
Gene Chromosome Location
Chr.5: 62306162-62387184 on Build GRCh38
Transcripts
NM_001098511.2 NM_001243952.1 NM_001243953.1 NM_004520.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
224280 c.742C>T p.R248C Chr.5: 62358250-62358250 on Build GRCh38
4653977 c.839T>C p.F280S Chr.5: 62358266-62358266 on Build GRCh38
1212453 c.724G>T p.D242Y Chr.5: 62358151-62358151 on Build GRCh38
4410424 c.673G>C p.D225H Chr.5: 62358181-62358181 on Build GRCh38
4188604 c.641A>G p.K214R Chr.5: 62358149-62358149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761341416 Chr.5: 62358309-62358309 on Build GRCh38
rs777689290 Chr.5: 62358195-62358195 on Build GRCh38
rs774218971 Chr.5: 62358302-62358302 on Build GRCh38
rs542499569 Chr.5: 62358311-62358311 on Build GRCh38
rs772229025 Chr.5: 62358186-62358186 on Build GRCh38
rs750414515 Chr.5: 62358111-62358111 on Build GRCh38
rs759996312 Chr.5: 62358241-62358241 on Build GRCh38
rs956243105 Chr.5: 62358243-62358243 on Build GRCh38
rs755115369 Chr.5: 62358317-62358318 on Build GRCh38
rs561322134 Chr.5: 62358140-62358140 on Build GRCh38
rs747164347 Chr.5: 62358287-62358287 on Build GRCh38
rs771593906 Chr.5: 62358204-62358204 on Build GRCh38
rs754709514 Chr.5: 62358144-62358144 on Build GRCh38
rs530240779 Chr.5: 62358279-62358279 on Build GRCh38
rs777214143 Chr.5: 62358231-62358231 on Build GRCh38
rs772621668 Chr.5: 62358321-62358321 on Build GRCh38
rs747058644 Chr.5: 62358197-62358197 on Build GRCh38
rs1002650078 Chr.5: 62358183-62358183 on Build GRCh38
rs748314027 Chr.5: 62358165-62358165 on Build GRCh38
rs779918646 Chr.5: 62358127-62358127 on Build GRCh38
rs779163986 Chr.5: 62358156-62358156 on Build GRCh38
rs1022678110 Chr.5: 62358113-62358113 on Build GRCh38

View Full Product Details