Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123784004-123784231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH10
Entrez Gene ID
196385
Gene Name
dynein axonemal heavy chain 10
Gene Aliases
-
UniGene
Hs.622654
Gene Chromosome Location
Chr.12: 123761995-123935720 on Build GRCh38
Transcripts
NM_207437.3 XM_005253555.3 XM_011538014.2 XM_011538015.2 XM_011538016.2 XM_011538017.2 XM_011538020.2 XM_017018960.1 XM_017018961.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1203933 c.1036C>T p.R346C Chr.12: 123784166-123784166 on Build GRCh38
1203934 c.490C>T p.R164C Chr.12: 123784166-123784166 on Build GRCh38
3457568 c.390C>T p.S130S Chr.12: 123784066-123784066 on Build GRCh38
4436485 c.912A>C p.K304N Chr.12: 123784042-123784042 on Build GRCh38
2226149 c.423G>A p.V141V Chr.12: 123784099-123784099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754794313 Chr.12: 123784157-123784157 on Build GRCh38
rs1007324776 Chr.12: 123784123-123784123 on Build GRCh38
rs922187155 Chr.12: 123784116-123784116 on Build GRCh38
rs780310767 Chr.12: 123784057-123784065 on Build GRCh38
rs771085939 Chr.12: 123784106-123784106 on Build GRCh38
rs754780592 Chr.12: 123784066-123784066 on Build GRCh38
rs150908267 Chr.12: 123784174-123784174 on Build GRCh38
rs1052725927 Chr.12: 123784044-123784044 on Build GRCh38
rs760223312 Chr.12: 123784136-123784136 on Build GRCh38
rs778755341 Chr.12: 123784069-123784069 on Build GRCh38
rs1051384753 Chr.12: 123784067-123784067 on Build GRCh38
rs776207660 Chr.12: 123784126-123784126 on Build GRCh38
rs369756810 Chr.12: 123784135-123784135 on Build GRCh38
rs764638695 Chr.12: 123784129-123784129 on Build GRCh38
rs749068543 Chr.12: 123784049-123784049 on Build GRCh38
rs758334070 Chr.12: 123784162-123784162 on Build GRCh38
rs969847214 Chr.12: 123784167-123784167 on Build GRCh38
rs752429146 Chr.12: 123784160-123784160 on Build GRCh38
rs377052101 Chr.12: 123784159-123784159 on Build GRCh38
rs777841997 Chr.12: 123784096-123784096 on Build GRCh38
rs747212453 Chr.12: 123784091-123784092 on Build GRCh38
rs747109313 Chr.12: 123784165-123784165 on Build GRCh38
rs377654704 Chr.12: 123784073-123784073 on Build GRCh38
rs772195311 Chr.12: 123784088-123784088 on Build GRCh38
rs201397099 Chr.12: 123784047-123784047 on Build GRCh38
rs550759010 Chr.12: 123784166-123784166 on Build GRCh38
rs141506707 Chr.12: 123784146-123784146 on Build GRCh38
rs373894230 Chr.12: 123784147-123784147 on Build GRCh38
rs750160351 Chr.12: 123784031-123784031 on Build GRCh38
rs767133369 Chr.12: 123784028-123784028 on Build GRCh38
rs777643907 Chr.12: 123784163-123784163 on Build GRCh38
rs758541072 Chr.12: 123784038-123784038 on Build GRCh38
rs776860831 Chr.12: 123784112-123784112 on Build GRCh38
rs148358039 Chr.12: 123784125-123784125 on Build GRCh38
rs759870580 Chr.12: 123784118-123784118 on Build GRCh38
rs763542837 Chr.12: 123784127-123784127 on Build GRCh38
rs903479165 Chr.12: 123784196-123784196 on Build GRCh38
rs547195870 Chr.12: 123784105-123784105 on Build GRCh38
rs755846507 Chr.12: 123784045-123784045 on Build GRCh38
rs770116938 Chr.12: 123784195-123784195 on Build GRCh38

View Full Product Details