Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76034650-76034919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ART3
Entrez Gene ID
419
Gene Name
ADP-ribosyltransferase 3
Gene Aliases
ARTC3
UniGene
Hs.24976
Gene Chromosome Location
Chr.4: 76011180-76112802 on Build GRCh38
Transcripts
NM_001130017.2 NM_001302123.1 NM_005409.4

Gene Symbol
CXCL11
Entrez Gene ID
6373
Gene Name
C-X-C motif chemokine ligand 11
Gene Aliases
H174, I-TAC, IP-9, IP9, SCYB11, SCYB9B, b-R1
UniGene
Hs.632592
Gene Chromosome Location
Chr.4: 76033682-76036197 on Build GRCh38
Transcripts
NM_001130017.2 NM_001302123.1 NM_005409.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4185414 c.268G>T p.E90* Chr.4: 76034810-76034810 on Build GRCh38
228397 c.272G>A p.R91K Chr.4: 76034806-76034806 on Build GRCh38
5734379 c.275_278delAGAA p.K92fs*>2 Chr.4: 76034800-76034803 on Build GRCh38
1431051 c.276G>T p.K92N Chr.4: 76034802-76034802 on Build GRCh38
6587246 c.265G>T p.V89F Chr.4: 76034813-76034813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537293787 Chr.4: 76034804-76034804 on Build GRCh38
rs6532111 Chr.4: 76034761-76034761 on Build GRCh38
rs746384125 Chr.4: 76034723-76034723 on Build GRCh38
rs773000689 Chr.4: 76034847-76034847 on Build GRCh38
rs757781879 Chr.4: 76034755-76034755 on Build GRCh38
rs932179284 Chr.4: 76034722-76034722 on Build GRCh38
rs540482675 Chr.4: 76034841-76034841 on Build GRCh38
rs764150862 Chr.4: 76034868-76034868 on Build GRCh38
rs752049858 Chr.4: 76034748-76034748 on Build GRCh38
rs779814819 Chr.4: 76034811-76034811 on Build GRCh38
rs748101280 Chr.4: 76034828-76034828 on Build GRCh38
rs746178066 Chr.4: 76034776-76034776 on Build GRCh38
rs186816745 Chr.4: 76034886-76034886 on Build GRCh38
rs367653764 Chr.4: 76034849-76034849 on Build GRCh38
rs748850682 Chr.4: 76034812-76034812 on Build GRCh38
rs773869103 Chr.4: 76034846-76034846 on Build GRCh38
rs904734397 Chr.4: 76034693-76034693 on Build GRCh38
rs370632418 Chr.4: 76034741-76034741 on Build GRCh38
rs764213946 Chr.4: 76034745-76034745 on Build GRCh38
rs944036125 Chr.4: 76034875-76034875 on Build GRCh38
rs61757197 Chr.4: 76034794-76034794 on Build GRCh38
rs775626155 Chr.4: 76034865-76034865 on Build GRCh38
rs565055838 Chr.4: 76034867-76034867 on Build GRCh38
rs765473239 Chr.4: 76034857-76034857 on Build GRCh38
rs773879571 Chr.4: 76034821-76034821 on Build GRCh38
rs980667331 Chr.4: 76034758-76034758 on Build GRCh38
rs768099347 Chr.4: 76034815-76034815 on Build GRCh38
rs952634518 Chr.4: 76034777-76034777 on Build GRCh38
rs1024734657 Chr.4: 76034738-76034738 on Build GRCh38
rs550670533 Chr.4: 76034895-76034895 on Build GRCh38

View Full Product Details