Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 50410896-50411135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMB
Entrez Gene ID
133418
Gene Name
embigin
Gene Aliases
GP70
UniGene
Hs.561411
Gene Chromosome Location
Chr.5: 50396192-50443299 on Build GRCh38
Transcripts
NM_198449.2 XM_011543146.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4547933 c.434G>C p.R145P Chr.5: 50410915-50410915 on Build GRCh38
1068540 c.444G>T p.K148N Chr.5: 50410905-50410905 on Build GRCh38
5403585 c.440A>C p.E147A Chr.5: 50410909-50410909 on Build GRCh38
3920142 c.429C>T p.F143F Chr.5: 50410920-50410920 on Build GRCh38
5605701 c.413G>A p.G138E Chr.5: 50410936-50410936 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549970108 Chr.5: 50411001-50411001 on Build GRCh38
rs773701425 Chr.5: 50411013-50411018 on Build GRCh38
rs752209701 Chr.5: 50410980-50410980 on Build GRCh38
rs775607646 Chr.5: 50411017-50411017 on Build GRCh38
rs1020442304 Chr.5: 50411044-50411044 on Build GRCh38
rs138980748 Chr.5: 50410958-50410958 on Build GRCh38
rs757848618 Chr.5: 50410991-50410991 on Build GRCh38
rs764624530 Chr.5: 50410922-50410922 on Build GRCh38
rs762212502 Chr.5: 50410936-50410936 on Build GRCh38
rs766063163 Chr.5: 50411028-50411031 on Build GRCh38
rs542972976 Chr.5: 50411073-50411073 on Build GRCh38
rs903280011 Chr.5: 50411047-50411047 on Build GRCh38
rs201037929 Chr.5: 50410961-50410961 on Build GRCh38
rs929700878 Chr.5: 50410946-50410946 on Build GRCh38
rs373289093 Chr.5: 50410994-50410994 on Build GRCh38
rs368346268 Chr.5: 50410933-50410933 on Build GRCh38
rs778481514 Chr.5: 50410978-50410978 on Build GRCh38
rs750195283 Chr.5: 50411000-50411000 on Build GRCh38
rs764759282 Chr.5: 50410938-50410938 on Build GRCh38
rs186844513 Chr.5: 50410995-50410995 on Build GRCh38
rs767981700 Chr.5: 50410940-50410940 on Build GRCh38
rs569361837 Chr.5: 50411066-50411066 on Build GRCh38
rs141564507 Chr.5: 50410965-50410965 on Build GRCh38
rs1033075729 Chr.5: 50411090-50411090 on Build GRCh38
rs756266996 Chr.5: 50411105-50411105 on Build GRCh38
rs745698101 Chr.5: 50410998-50410998 on Build GRCh38

View Full Product Details