Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84868750-84869023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISPLD2
Entrez Gene ID
83716
Gene Name
cysteine rich secretory protein LCCL domain containing 2
Gene Aliases
CRISP11, LCRISP2
UniGene
Hs.513779
Gene Chromosome Location
Chr.16: 84819981-84909510 on Build GRCh38
Transcripts
NM_031476.3 XM_005256190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
974478 c.854-1G>T p.? Chr.16: 84868850-84868850 on Build GRCh38
6277243 c.905C>T p.T302M Chr.16: 84868902-84868902 on Build GRCh38
6277244 c.905C>T p.T302M Chr.16: 84868902-84868902 on Build GRCh38
1661279 c.862G>A p.V288I Chr.16: 84868859-84868859 on Build GRCh38
6635549 c.906G>A p.T302T Chr.16: 84868903-84868903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549424946 Chr.16: 84868951-84868951 on Build GRCh38
rs760238602 Chr.16: 84868918-84868953 on Build GRCh38
rs774983701 Chr.16: 84868913-84868948 on Build GRCh38
rs376476534 Chr.16: 84868822-84868822 on Build GRCh38
rs965770604 Chr.16: 84868794-84868794 on Build GRCh38
rs373850084 Chr.16: 84868937-84868937 on Build GRCh38
rs370925447 Chr.16: 84868948-84868948 on Build GRCh38
rs780093987 Chr.16: 84868963-84868963 on Build GRCh38
rs369419356 Chr.16: 84868821-84868821 on Build GRCh38
rs774119535 Chr.16: 84868939-84868939 on Build GRCh38
rs754226615 Chr.16: 84868945-84868945 on Build GRCh38
rs376411279 Chr.16: 84868805-84868805 on Build GRCh38
rs897738059 Chr.16: 84868846-84868846 on Build GRCh38
rs78626142 Chr.16: 84868950-84868950 on Build GRCh38
rs997287509 Chr.16: 84868798-84868798 on Build GRCh38
rs770919401 Chr.16: 84868903-84868903 on Build GRCh38
rs371310544 Chr.16: 84868844-84868844 on Build GRCh38
rs752855933 Chr.16: 84868882-84868882 on Build GRCh38
rs8045364 Chr.16: 84868935-84868935 on Build GRCh38
rs766721884 Chr.16: 84868859-84868859 on Build GRCh38
rs915510587 Chr.16: 84868982-84868982 on Build GRCh38
rs993443912 Chr.16: 84868852-84868852 on Build GRCh38
rs745603203 Chr.16: 84868929-84868929 on Build GRCh38
rs747188693 Chr.16: 84868902-84868902 on Build GRCh38
rs3840043 Chr.16: 84868951-84868986 on Build GRCh38
rs769767232 Chr.16: 84868827-84868827 on Build GRCh38
rs796487 Chr.16: 84868943-84868943 on Build GRCh38
rs778133931 Chr.16: 84868932-84868932 on Build GRCh38
rs370247748 Chr.16: 84868956-84868956 on Build GRCh38
rs1004829369 Chr.16: 84868975-84868975 on Build GRCh38
rs772363922 Chr.16: 84868820-84868820 on Build GRCh38
rs769881616 Chr.16: 84868931-84868931 on Build GRCh38
rs944473176 Chr.16: 84868992-84868992 on Build GRCh38
rs759544076 Chr.16: 84868836-84868836 on Build GRCh38
rs771797609 Chr.16: 84868854-84868855 on Build GRCh38
rs533922531 Chr.16: 84868990-84868990 on Build GRCh38
rs997235638 Chr.16: 84869000-84869000 on Build GRCh38
rs779014549 Chr.16: 84868835-84868835 on Build GRCh38
rs781141782 Chr.16: 84868908-84868908 on Build GRCh38
rs977380309 Chr.16: 84868904-84868904 on Build GRCh38
rs762328714 Chr.16: 84868837-84868837 on Build GRCh38
rs560521019 Chr.16: 84868946-84868946 on Build GRCh38
rs1052543499 Chr.16: 84868787-84868787 on Build GRCh38
rs778965515 Chr.16: 84868818-84868818 on Build GRCh38
rs778110083 Chr.16: 84868887-84868887 on Build GRCh38
rs777714027 Chr.16: 84868952-84868952 on Build GRCh38
rs201418783 Chr.16: 84868869-84868869 on Build GRCh38
rs74934991 Chr.16: 84868968-84868968 on Build GRCh38
rs1024105519 Chr.16: 84868886-84868886 on Build GRCh38
rs763591003 Chr.16: 84868936-84868971 on Build GRCh38
rs756555506 Chr.16: 84868858-84868858 on Build GRCh38
rs780583267 Chr.16: 84868815-84868815 on Build GRCh38
rs202056245 Chr.16: 84868953-84868953 on Build GRCh38
rs769686560 Chr.16: 84868961-84868961 on Build GRCh38
rs117748842 Chr.16: 84868816-84868816 on Build GRCh38
rs770609688 Chr.16: 84868828-84868828 on Build GRCh38
rs946842707 Chr.16: 84868987-84868987 on Build GRCh38
rs768038146 Chr.16: 84868938-84868938 on Build GRCh38
rs969445457 Chr.16: 84868888-84868888 on Build GRCh38
rs575567977 Chr.16: 84868819-84868819 on Build GRCh38
rs757570103 Chr.16: 84868954-84868954 on Build GRCh38
rs761376049 Chr.16: 84868940-84868940 on Build GRCh38
rs879066816 Chr.16: 84868941-84868941 on Build GRCh38
rs756402664 Chr.16: 84868809-84868809 on Build GRCh38
rs72799568 Chr.16: 84868877-84868877 on Build GRCh38
rs745562352 Chr.16: 84868989-84868989 on Build GRCh38
rs1004546363 Chr.16: 84868784-84868784 on Build GRCh38
rs765774079 Chr.16: 84868947-84868947 on Build GRCh38
rs866428176 Chr.16: 84868874-84868874 on Build GRCh38
rs148934412 Chr.16: 84868866-84868866 on Build GRCh38
rs1014619139 Chr.16: 84868785-84868785 on Build GRCh38
rs537819550 Chr.16: 84868955-84868955 on Build GRCh38
rs763846881 Chr.16: 84868842-84868842 on Build GRCh38

View Full Product Details