Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7689340-7689575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCER2
Entrez Gene ID
2208
Gene Name
Fc fragment of IgE receptor II
Gene Aliases
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF
UniGene
Hs.465778
Gene Chromosome Location
Chr.19: 7688757-7702755 on Build GRCh38
Transcripts
NM_001207019.2 NM_001220500.1 NM_002002.4 XM_005272462.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3371557 c.740C>T p.P247L Chr.19: 7689419-7689419 on Build GRCh38
1189810 c.785T>C p.M262T Chr.19: 7689374-7689374 on Build GRCh38
6662663 c.819C>T p.C273C Chr.19: 7689340-7689340 on Build GRCh38
7367984 c.745G>A p.E249K Chr.19: 7689414-7689414 on Build GRCh38
1397652 c.788G>A p.R263Q Chr.19: 7689371-7689371 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973200988 Chr.19: 7689556-7689556 on Build GRCh38
rs199808473 Chr.19: 7689401-7689401 on Build GRCh38
rs933008811 Chr.19: 7689366-7689366 on Build GRCh38
rs1048333794 Chr.19: 7689545-7689545 on Build GRCh38
rs181728660 Chr.19: 7689555-7689555 on Build GRCh38
rs770191728 Chr.19: 7689407-7689407 on Build GRCh38
rs116058720 Chr.19: 7689448-7689448 on Build GRCh38
rs753517266 Chr.19: 7689457-7689457 on Build GRCh38
rs534559698 Chr.19: 7689466-7689466 on Build GRCh38
rs1002333028 Chr.19: 7689424-7689424 on Build GRCh38
rs778075952 Chr.19: 7689471-7689471 on Build GRCh38
rs933735344 Chr.19: 7689418-7689418 on Build GRCh38
rs910274311 Chr.19: 7689487-7689487 on Build GRCh38
rs777512868 Chr.19: 7689371-7689371 on Build GRCh38
rs760297884 Chr.19: 7689429-7689429 on Build GRCh38
rs932517496 Chr.19: 7689529-7689529 on Build GRCh38
rs753680245 Chr.19: 7689374-7689374 on Build GRCh38
rs771315020 Chr.19: 7689475-7689475 on Build GRCh38
rs555446476 Chr.19: 7689422-7689422 on Build GRCh38
rs755110246 Chr.19: 7689425-7689426 on Build GRCh38
rs767049788 Chr.19: 7689423-7689423 on Build GRCh38
rs913968252 Chr.19: 7689469-7689471 on Build GRCh38
rs774226662 Chr.19: 7689489-7689489 on Build GRCh38
rs749193919 Chr.19: 7689381-7689391 on Build GRCh38
rs555861824 Chr.19: 7689465-7689465 on Build GRCh38
rs774864408 Chr.19: 7689414-7689414 on Build GRCh38
rs748192150 Chr.19: 7689468-7689468 on Build GRCh38
rs778583417 Chr.19: 7689464-7689464 on Build GRCh38
rs200736793 Chr.19: 7689408-7689408 on Build GRCh38
rs558024931 Chr.19: 7689474-7689474 on Build GRCh38
rs548023601 Chr.19: 7689359-7689359 on Build GRCh38
rs954323464 Chr.19: 7689473-7689473 on Build GRCh38
rs771204560 Chr.19: 7689412-7689412 on Build GRCh38
rs377209516 Chr.19: 7689399-7689399 on Build GRCh38
rs752243016 Chr.19: 7689427-7689427 on Build GRCh38
rs138745804 Chr.19: 7689381-7689381 on Build GRCh38
rs757006186 Chr.19: 7689463-7689463 on Build GRCh38
rs370424564 Chr.19: 7689364-7689364 on Build GRCh38
rs1036781021 Chr.19: 7689382-7689382 on Build GRCh38
rs1033691014 Chr.19: 7689433-7689433 on Build GRCh38
rs1011049136 Chr.19: 7689461-7689461 on Build GRCh38
rs941811076 Chr.19: 7689391-7689391 on Build GRCh38
rs779075233 Chr.19: 7689444-7689445 on Build GRCh38
rs201945831 Chr.19: 7689402-7689402 on Build GRCh38
rs117396061 Chr.19: 7689365-7689365 on Build GRCh38
rs35825847 Chr.19: 7689398-7689398 on Build GRCh38
rs752039664 Chr.19: 7689360-7689360 on Build GRCh38
rs770026851 Chr.19: 7689469-7689469 on Build GRCh38
rs201361607 Chr.19: 7689363-7689363 on Build GRCh38

View Full Product Details