Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 96880130-96880355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERAP1
Entrez Gene ID
51752
Gene Name
endoplasmic reticulum aminopeptidase 1
Gene Aliases
A-LAP, ALAP, APPILS, ARTS-1, ARTS1, ERAAP, ERAAP1, PILS-AP, PILSAP
UniGene
Hs.666524
Gene Chromosome Location
Chr.5: 96759245-96935983 on Build GRCh38
Transcripts
XM_011543480.2 XM_011543481.2 XM_011543484.2 XM_011543485.2 XM_011543486.2 XM_017009580.1 XM_017009581.1 XM_011543544.1

Gene Symbol
ERAP2
Entrez Gene ID
64167
Gene Name
endoplasmic reticulum aminopeptidase 2
Gene Aliases
L-RAP, LRAP
UniGene
Hs.482910
Gene Chromosome Location
Chr.5: 96875940-96919703 on Build GRCh38
Transcripts
XM_011543480.2 XM_011543481.2 XM_011543484.2 XM_011543485.2 XM_011543486.2 XM_017009580.1 XM_017009581.1 XM_011543544.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6316517 c.524G>T p.G175V Chr.5: 96880209-96880209 on Build GRCh38
5472135 c.470C>T p.T157M Chr.5: 96880155-96880155 on Build GRCh38
5457211 c.514T>G p.L172V Chr.5: 96880199-96880199 on Build GRCh38
4895542 c.525C>T p.G175G Chr.5: 96880210-96880210 on Build GRCh38
6895171 c.565G>T p.G189C Chr.5: 96880250-96880250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533852348 Chr.5: 96880230-96880230 on Build GRCh38
rs779492267 Chr.5: 96880195-96880195 on Build GRCh38
rs778597546 Chr.5: 96880305-96880306 on Build GRCh38
rs747756184 Chr.5: 96880235-96880235 on Build GRCh38
rs372960918 Chr.5: 96880271-96880271 on Build GRCh38
rs781565196 Chr.5: 96880158-96880158 on Build GRCh38
rs770883922 Chr.5: 96880270-96880270 on Build GRCh38
rs757591290 Chr.5: 96880157-96880157 on Build GRCh38
rs751958377 Chr.5: 96880156-96880156 on Build GRCh38
rs201186013 Chr.5: 96880310-96880310 on Build GRCh38
rs780640903 Chr.5: 96880155-96880155 on Build GRCh38
rs772989785 Chr.5: 96880241-96880241 on Build GRCh38
rs762988047 Chr.5: 96880276-96880276 on Build GRCh38
rs778740282 Chr.5: 96880229-96880229 on Build GRCh38
rs1004726355 Chr.5: 96880174-96880174 on Build GRCh38
rs754623786 Chr.5: 96880303-96880305 on Build GRCh38
rs750786882 Chr.5: 96880309-96880309 on Build GRCh38
rs764116922 Chr.5: 96880287-96880287 on Build GRCh38
rs978384778 Chr.5: 96880243-96880243 on Build GRCh38
rs142952967 Chr.5: 96880289-96880289 on Build GRCh38
rs767837857 Chr.5: 96880301-96880301 on Build GRCh38
rs564736676 Chr.5: 96880218-96880218 on Build GRCh38
rs762029913 Chr.5: 96880292-96880292 on Build GRCh38
rs989270158 Chr.5: 96880180-96880180 on Build GRCh38
rs964548900 Chr.5: 96880177-96880177 on Build GRCh38
rs140628405 Chr.5: 96880209-96880209 on Build GRCh38
rs750901853 Chr.5: 96880160-96880160 on Build GRCh38
rs563004439 Chr.5: 96880298-96880298 on Build GRCh38
rs756643881 Chr.5: 96880187-96880187 on Build GRCh38
rs558767630 Chr.5: 96880252-96880252 on Build GRCh38
rs552083216 Chr.5: 96880207-96880207 on Build GRCh38

View Full Product Details