Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57516666-57516939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDIT3
Entrez Gene ID
1649
Gene Name
DNA damage inducible transcript 3
Gene Aliases
CEBPZ, CHOP, CHOP-10, CHOP10, GADD153
UniGene
Hs.505777
Gene Chromosome Location
Chr.12: 57516588-57520517 on Build GRCh38
Transcripts
NM_001195053.1 NM_001195054.1 NM_001195055.1 NM_001195056.1 NM_001195057.1 NM_004083.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5361545 c.530C>T p.A177V Chr.12: 57516858-57516858 on Build GRCh38
942078 c.435C>T p.I145I Chr.12: 57516884-57516884 on Build GRCh38
5451270 c.504C>G p.I168M Chr.12: 57516884-57516884 on Build GRCh38
942077 c.440G>A p.R147H Chr.12: 57516879-57516879 on Build GRCh38
6641334 c.418C>T p.R140W Chr.12: 57516901-57516901 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776822428 Chr.12: 57516852-57516852 on Build GRCh38
rs1057229054 Chr.12: 57516812-57516812 on Build GRCh38
rs182097761 Chr.12: 57516716-57516716 on Build GRCh38
rs377303534 Chr.12: 57516900-57516900 on Build GRCh38
rs762234408 Chr.12: 57516853-57516853 on Build GRCh38
rs775586763 Chr.12: 57516834-57516834 on Build GRCh38
rs981054561 Chr.12: 57516817-57516817 on Build GRCh38
rs529041005 Chr.12: 57516698-57516698 on Build GRCh38
rs971306627 Chr.12: 57516805-57516805 on Build GRCh38
rs897889899 Chr.12: 57516697-57516697 on Build GRCh38
rs777269541 Chr.12: 57516797-57516804 on Build GRCh38
rs924335571 Chr.12: 57516836-57516836 on Build GRCh38
rs921111628 Chr.12: 57516734-57516735 on Build GRCh38
rs897862749 Chr.12: 57516895-57516895 on Build GRCh38
rs568713877 Chr.12: 57516858-57516858 on Build GRCh38
rs747778183 Chr.12: 57516779-57516779 on Build GRCh38
rs745766747 Chr.12: 57516804-57516804 on Build GRCh38
rs765557061 Chr.12: 57516857-57516857 on Build GRCh38
rs745810892 Chr.12: 57516917-57516917 on Build GRCh38
rs946840113 Chr.12: 57516869-57516869 on Build GRCh38
rs1027897283 Chr.12: 57516741-57516741 on Build GRCh38
rs905377922 Chr.12: 57516870-57516870 on Build GRCh38
rs766968503 Chr.12: 57516878-57516878 on Build GRCh38
rs993628420 Chr.12: 57516709-57516709 on Build GRCh38
rs561733924 Chr.12: 57516694-57516694 on Build GRCh38
rs878950898 Chr.12: 57516757-57516757 on Build GRCh38
rs563855250 Chr.12: 57516739-57516739 on Build GRCh38
rs770436974 Chr.12: 57516906-57516939 on Build GRCh38
rs975219878 Chr.12: 57516732-57516733 on Build GRCh38
rs780733767 Chr.12: 57516767-57516767 on Build GRCh38
rs778826051 Chr.12: 57516908-57516908 on Build GRCh38
rs141272899 Chr.12: 57516849-57516849 on Build GRCh38
rs200022707 Chr.12: 57516884-57516884 on Build GRCh38
rs758889844 Chr.12: 57516871-57516871 on Build GRCh38
rs368301051 Chr.12: 57516786-57516786 on Build GRCh38
rs11547020 Chr.12: 57516765-57516765 on Build GRCh38
rs755753113 Chr.12: 57516783-57516783 on Build GRCh38
rs1020049484 Chr.12: 57516901-57516901 on Build GRCh38
rs150793545 Chr.12: 57516866-57516866 on Build GRCh38
rs772089247 Chr.12: 57516807-57516807 on Build GRCh38
rs748594689 Chr.12: 57516843-57516844 on Build GRCh38
rs965875703 Chr.12: 57516723-57516723 on Build GRCh38
rs748943913 Chr.12: 57516887-57516887 on Build GRCh38
rs139206730 Chr.12: 57516879-57516879 on Build GRCh38
rs774493745 Chr.12: 57516798-57516798 on Build GRCh38
rs112917609 Chr.12: 57516883-57516883 on Build GRCh38
rs754669482 Chr.12: 57516850-57516850 on Build GRCh38
rs1022807484 Chr.12: 57516800-57516800 on Build GRCh38

View Full Product Details