Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74531224-74531493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTRA2
Entrez Gene ID
27429
Gene Name
HtrA serine peptidase 2
Gene Aliases
OMI, PARK13, PRSS25
UniGene
Hs.469045
Gene Chromosome Location
Chr.2: 74529405-74533556 on Build GRCh38
Transcripts
NM_001321727.1 NM_001321728.1 NM_013247.4 NM_145074.2 NR_135769.1 NR_135770.1 NR_135771.1 NR_135772.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7313026 c.907-7C>T p.? Chr.2: 74531332-74531332 on Build GRCh38
4562095 c.911G>A p.G304E Chr.2: 74531343-74531343 on Build GRCh38
2999887 c.935A>T p.N312I Chr.2: 74531367-74531367 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs34239071 Chr.2: 74531307-74531308 on Build GRCh38
rs768956001 Chr.2: 74531317-74531317 on Build GRCh38
rs766035905 Chr.2: 74531369-74531369 on Build GRCh38
rs780218548 Chr.2: 74531309-74531309 on Build GRCh38
rs533292599 Chr.2: 74531436-74531436 on Build GRCh38
rs770994075 Chr.2: 74531459-74531460 on Build GRCh38
rs370645837 Chr.2: 74531332-74531332 on Build GRCh38
rs886065463 Chr.2: 74531323-74531323 on Build GRCh38
rs772596976 Chr.2: 74531465-74531465 on Build GRCh38
rs757381657 Chr.2: 74531288-74531288 on Build GRCh38
rs369607927 Chr.2: 74531290-74531290 on Build GRCh38
rs1043056207 Chr.2: 74531460-74531460 on Build GRCh38
rs759371209 Chr.2: 74531396-74531396 on Build GRCh38
rs774913561 Chr.2: 74531318-74531318 on Build GRCh38
rs371349095 Chr.2: 74531267-74531267 on Build GRCh38
rs116244977 Chr.2: 74531426-74531426 on Build GRCh38
rs1046251279 Chr.2: 74531401-74531401 on Build GRCh38
rs779195139 Chr.2: 74531450-74531450 on Build GRCh38
rs750538240 Chr.2: 74531427-74531427 on Build GRCh38
rs760279402 Chr.2: 74531333-74531333 on Build GRCh38
rs769029206 Chr.2: 74531444-74531444 on Build GRCh38
rs764980920 Chr.2: 74531413-74531413 on Build GRCh38
rs749674762 Chr.2: 74531312-74531312 on Build GRCh38
rs373380407 Chr.2: 74531386-74531386 on Build GRCh38
rs943659158 Chr.2: 74531249-74531249 on Build GRCh38
rs374020540 Chr.2: 74531302-74531302 on Build GRCh38
rs780359261 Chr.2: 74531322-74531322 on Build GRCh38
rs750429164 Chr.2: 74531269-74531269 on Build GRCh38
rs529245240 Chr.2: 74531295-74531295 on Build GRCh38
rs752661709 Chr.2: 74531425-74531425 on Build GRCh38
rs201936888 Chr.2: 74531320-74531320 on Build GRCh38
rs748604002 Chr.2: 74531452-74531452 on Build GRCh38
rs756344442 Chr.2: 74531430-74531430 on Build GRCh38

View Full Product Details