Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73105182-73105451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNQ5
Entrez Gene ID
56479
Gene Name
potassium voltage-gated channel subfamily Q member 5
Gene Aliases
Kv7.5
UniGene
Hs.445324
Gene Chromosome Location
Chr.6: 72621843-73198851 on Build GRCh38
Transcripts
NM_001160130.1 NM_001160132.1 NM_001160133.1 NM_001160134.1 NM_019842.3 XM_017011058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5835257 c.1020A>C p.A340A Chr.6: 73105358-73105358 on Build GRCh38
5830507 c.976T>C p.L326L Chr.6: 73105314-73105314 on Build GRCh38
743179 c.958C>A p.L320I Chr.6: 73105296-73105296 on Build GRCh38
6241111 c.978G>T p.L326F Chr.6: 73105316-73105316 on Build GRCh38
7309280 c.994G>T p.A332S Chr.6: 73105332-73105332 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753118346 Chr.6: 73105331-73105331 on Build GRCh38
rs767953445 Chr.6: 73105314-73105314 on Build GRCh38
rs754372915 Chr.6: 73105370-73105370 on Build GRCh38
rs774031694 Chr.6: 73105301-73105301 on Build GRCh38
rs764651477 Chr.6: 73105367-73105367 on Build GRCh38
rs77753084 Chr.6: 73105397-73105397 on Build GRCh38
rs758619904 Chr.6: 73105413-73105413 on Build GRCh38
rs780992183 Chr.6: 73105235-73105235 on Build GRCh38
rs140427666 Chr.6: 73105426-73105426 on Build GRCh38
rs770386902 Chr.6: 73105286-73105286 on Build GRCh38
rs376842647 Chr.6: 73105249-73105249 on Build GRCh38
rs773079348 Chr.6: 73105262-73105262 on Build GRCh38
rs778912698 Chr.6: 73105398-73105398 on Build GRCh38
rs780904289 Chr.6: 73105415-73105415 on Build GRCh38
rs762517911 Chr.6: 73105263-73105263 on Build GRCh38
rs757343190 Chr.6: 73105215-73105215 on Build GRCh38
rs540453351 Chr.6: 73105220-73105220 on Build GRCh38
rs912621198 Chr.6: 73105420-73105420 on Build GRCh38
rs768421041 Chr.6: 73105229-73105229 on Build GRCh38
rs138701359 Chr.6: 73105310-73105310 on Build GRCh38
rs750615489 Chr.6: 73105406-73105406 on Build GRCh38
rs748008833 Chr.6: 73105240-73105240 on Build GRCh38
rs753967154 Chr.6: 73105208-73105208 on Build GRCh38
rs779194264 Chr.6: 73105217-73105217 on Build GRCh38
rs761207241 Chr.6: 73105334-73105334 on Build GRCh38

View Full Product Details