Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94494745-94495002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1429
Entrez Gene ID
25962
Gene Name
KIAA1429
Gene Aliases
MSTP054, fSAP121
UniGene
Hs.202238
Gene Chromosome Location
Chr.8: 94487777-94553518 on Build GRCh38
Transcripts
NM_015496.4 XM_017013306.1 XM_017013307.1 XM_017013308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1643661 c.4575T>C p.D1525D Chr.8: 94494926-94494926 on Build GRCh38
5803270 c.4545-1G>A p.? Chr.8: 94494957-94494957 on Build GRCh38
3925721 c.4604C>T p.P1535L Chr.8: 94494897-94494897 on Build GRCh38
4695647 c.4570A>G p.M1524V Chr.8: 94494931-94494931 on Build GRCh38
361057 c.4629A>G p.T1543T Chr.8: 94494872-94494872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552011929 Chr.8: 94494827-94494827 on Build GRCh38
rs138979789 Chr.8: 94494946-94494946 on Build GRCh38
rs777019768 Chr.8: 94494900-94494900 on Build GRCh38
rs778043822 Chr.8: 94494829-94494829 on Build GRCh38
rs565884979 Chr.8: 94494829-94494829 on Build GRCh38
rs893460375 Chr.8: 94494794-94494794 on Build GRCh38
rs758768324 Chr.8: 94494822-94494822 on Build GRCh38
rs758817547 Chr.8: 94494834-94494834 on Build GRCh38
rs763006732 Chr.8: 94494948-94494948 on Build GRCh38
rs752974984 Chr.8: 94494820-94494822 on Build GRCh38
rs747059214 Chr.8: 94494844-94494844 on Build GRCh38
rs748361563 Chr.8: 94494928-94494928 on Build GRCh38
rs376087934 Chr.8: 94494966-94494966 on Build GRCh38
rs186921537 Chr.8: 94494803-94494803 on Build GRCh38
rs1007631421 Chr.8: 94494881-94494881 on Build GRCh38
rs963372947 Chr.8: 94494932-94494932 on Build GRCh38
rs146145980 Chr.8: 94494910-94494910 on Build GRCh38
rs376923485 Chr.8: 94494828-94494828 on Build GRCh38
rs972009796 Chr.8: 94494959-94494959 on Build GRCh38
rs73274021 Chr.8: 94494832-94494832 on Build GRCh38
rs1053396313 Chr.8: 94494786-94494786 on Build GRCh38
rs112174315 Chr.8: 94494970-94494970 on Build GRCh38
rs1003663006 Chr.8: 94494893-94494893 on Build GRCh38
rs772881638 Chr.8: 94494947-94494947 on Build GRCh38
rs139312864 Chr.8: 94494793-94494793 on Build GRCh38
rs752306755 Chr.8: 94494813-94494813 on Build GRCh38
rs1016299850 Chr.8: 94494905-94494905 on Build GRCh38
rs199895737 Chr.8: 94494843-94494843 on Build GRCh38
rs768813160 Chr.8: 94494873-94494873 on Build GRCh38

View Full Product Details